Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EFCAB6 SIRNA Assays ›
Gene Symbol:
EFCAB6
Gene Name:
EF-hand calcium binding domain 6
Gene Aliases:
DJBP, HSCBCIP1, dJ185D5.1
Chromosome Location:
Chr.22: 43528549 - 43812403 on Build GRCh38
UniGene ID:
Hs.658996
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
30390
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EFCAB6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022785.3
NP_073622.2
26
3666
NM_198856.2
NP_942153.1
24
3308
XM_005261704.2
XP_005261761.1
26
3606
XM_005261705.3
XP_005261762.1
25
3520
XM_011530316.1
XP_011528618.1
27
3893
XM_011530317.2
XP_011528619.1
25
3453
XM_011530318.2
XP_011528620.1
25
3549
XM_011530319.2
XP_011528621.1
24
3241
XM_011530320.2
XP_011528622.1
27
3705
XM_011530321.2
XP_011528623.1
24
3236
XM_011530322.2
XP_011528624.1
25
3364
XM_011530323.2
XP_011528625.1
22
3022
XM_011530325.2
XP_011528627.1
20
2767
XM_011530326.2
XP_011528628.1
27
3705
XM_011530327.2
XP_011528629.1
17
2514
XM_011530329.1
XP_011528631.1
13
1986
XM_017028910.1
XP_016884399.1
27
3705
XM_017028911.1
XP_016884400.1
27
3705
XM_017028912.1
XP_016884401.1
23
3364
GenBank mRNA
AB051459.1
-
12
1849
AB073862.1
-
6
860
AK027241.1
-
6
864
AK225596.1
-
6
865
AK225605.1
-
6
865
AK292411.1
-
26
3606
AK292498.1
-
24
3285
AK309013.1
-
7
1064
BC039315.1
-
24
3161
5297436
BP325901.1
-
2
206
Back To Top
More Information
Panther Classification:
Molecular Function -
calmodulin-related
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-rxbn5:80/10.239.162.255:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0