Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ATRX SIRNA Assays ›
          Gene Symbol:
          ATRX
          Gene Name:
          ATRX, chromatin remodeler
          Gene Aliases:
          ATR2, JMS, MRX52, MRXHF1, RAD54, RAD54L, SFM1, SHS, XH2, XNP, ZNF-HX
          Chromosome Location:
          Chr.X: 77504878 - 77786269 on Build GRCh38
          UniGene ID:
          Hs.533526
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 3075

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ATRX
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000489.4 NP_000480.3 9 952
          NM_138270.3 NP_612114.2 8 838
          XM_005262153.4 XP_005262210.2 9 949
          XM_005262154.4 XP_005262211.2 9 952
          XM_005262155.4 XP_005262212.2 9 1053
          XM_005262156.4 XP_005262213.2 8 805
          XM_005262157.4 XP_005262214.2 8 835
          XM_006724666.3 XP_006724729.1 8 835
          XM_006724667.3 XP_006724730.1 7 691
          XM_006724668.2 XP_006724731.1 9 952
          XM_017029601.1 XP_016885090.1 9 949
          XM_017029602.1 XP_016885091.1 9 1033
          XM_017029603.1 XP_016885092.1 8 802
          XM_017029604.1 XP_016885093.1 8 838
          XM_017029605.1 XP_016885094.1 9 1053
          XM_017029606.1 XP_016885095.1 8 939
          XM_017029607.1 XP_016885096.1 8 936
          XM_017029608.1 XP_016885097.1 7 688
          XM_017029609.1 XP_016885098.1 8 939
          XM_017029610.1 XP_016885099.1 8 936
          XM_017029611.1 XP_016885100.1 7 691
          GenBank mRNA AB102641.1 - 9 684
          AB208928.1 - 8 766
          AK307791.1 - 8 778
          U72936.2 - 10 1021
          U72937.2 - 9 899
          U72938.2 - 8 785

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          DNA methylation
          DNA recombination
          nucleosome assembly
          DNA replication-independent nucleosome assembly
          chromatin remodeling
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          spermatogenesis
          positive regulation of nuclear cell cycle DNA replication
          covalent chromatin modification
          DNA damage response, signal transduction by p53 class mediator
          forebrain development
          replication fork processing
          positive regulation of telomere maintenance
          DNA duplex unwinding
          post-embryonic forelimb morphogenesis
          multicellular organism growth
          positive regulation of transcription from RNA polymerase II promoter
          Sertoli cell development
          protein localization to chromosome, telomeric region
          seminiferous tubule development
          cellular response to hydroxyurea
          regulation of histone H3-K9 trimethylation
          negative regulation of telomeric RNA transcription from RNA pol II promoter
          positive regulation of telomeric RNA transcription from RNA pol II promoter
          negative regulation of maintenance of mitotic sister chromatid cohesion, telomeric
          DNA binding
          DNA helicase activity
          chromatin binding
          helicase activity
          protein binding
          ATP binding
          DNA translocase activity
          methylated histone binding
          histone binding
          metal ion binding
          chromo shadow domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0