Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CARF SIRNA Assays ›
Gene Symbol:
CARF
Gene Name:
calcium responsive transcription factor
Gene Aliases:
ALS2CR8, NYD-SP24
Chromosome Location:
Chr.2: 202912218 - 202987856 on Build GRCh38
UniGene ID:
Hs.444982
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
31723
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CARF
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001104586.2
NP_001098056.1
16
2558
NM_001282910.1
NP_001269839.1
15
2330
NM_001282911.1
NP_001269840.1
16
2411
NM_001282912.1
NP_001269841.1
14
2209
NM_001322427.1
NP_001309356.1
17
2617
NM_001322428.1
NP_001309357.1
16
2831
NM_001322429.1
NP_001309358.1
15
2437
NM_001322430.1
NP_001309359.1
14
1998
NM_024744.15
NP_079020.13
17
3455
NR_136330.1
15
2437
NR_136331.1
14
1998
XM_005246858.3
XP_005246915.1
17
2564
XM_005246859.3
XP_005246916.1
17
2616
XM_005246864.3
XP_005246921.1
9
1490
XM_011511867.2
XP_011510169.1
17
2595
XM_011511868.2
XP_011510170.1
15
2129
XM_011511869.2
XP_011510171.1
9
1506
XM_017004961.1
XP_016860450.1
16
2953
XM_017004962.1
XP_016860451.1
14
2185
XM_017004963.1
XP_016860452.1
15
2253
XM_017004964.1
XP_016860453.1
14
2163
XM_017004965.1
XP_016860454.1
13
1971
XM_017004966.1
XP_016860455.1
12
1678
GenBank mRNA
AB053309.1
-
17
2936
AF454946.1
-
14
2285
AK025232.1
-
13
2158
AK225218.1
-
13
2158
AK304014.1
-
15
2265
AK316432.1
-
16
2346
AY032876.2
-
16
2510
BC033777.2
-
16
2571
4521845
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
cellular response to potassium ion
positive regulation of transcription from RNA polymerase II promoter in response to calcium ion
cellular response to calcium ion
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0