Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZRANB3 SIRNA Assays ›
          Gene Symbol:
          ZRANB3
          Gene Name:
          zinc finger RANBP2-type containing 3
          Gene Aliases:
          4933425L19Rik, AH2
          Chromosome Location:
          Chr.2: 135164703 - 135531322 on Build GRCh38
          UniGene ID:
          Hs.658422
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 33820

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZRANB3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001286568.1 NP_001273497.1 14 2206
          NM_001286569.1 NP_001273498.1 15 2329
          NM_032143.3 NP_115519.2 14 2111
          XM_005263809.1 XP_005263866.1 14 2065
          XM_006712788.1 XP_006712851.1 14 2039
          XM_011511958.2 XP_011510260.1 14 2554
          XM_011511959.2 XP_011510261.2 14 2119
          XM_011511960.1 XP_011510262.1 14 2087
          XM_011511961.2 XP_011510263.2 14 2053
          XM_011511963.1 XP_011510265.1 14 2058
          XM_011511966.2 XP_011510268.2 12 1937
          XM_017005065.1 XP_016860554.1 14 2058
          XM_017005066.1 XP_016860555.1 14 2117
          XM_017005067.1 XP_016860556.1 14 2065
          XM_017005068.1 XP_016860557.1 14 2039
          XM_017005069.1 XP_016860558.1 14 2087
          XM_017005070.1 XP_016860559.1 14 2087
          XM_017005071.1 XP_016860560.1 12 1858
          GenBank mRNA AK095362.1 - 13 1769
          AK131303.1 - 8 1255
          AK304601.1 - 2 446
          AL136824.1 - 3 553
          AL833322.1 - 15 2210
          BC089429.1 - 14 2111 3077975
          BX647838.1 - 14 2234

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA strand renaturation
          DNA repair
          cellular response to DNA damage stimulus
          response to UV
          replication fork processing
          DNA rewinding
          negative regulation of DNA recombination
          replication fork protection
          nucleic acid phosphodiester bond hydrolysis
          DNA helicase activity
          endodeoxyribonuclease activity
          protein binding
          ATP binding
          zinc ion binding
          annealing helicase activity
          K63-linked polyubiquitin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0