Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ASCC2 SIRNA Assays ›
          Gene Symbol:
          ASCC2
          Gene Name:
          activating signal cointegrator 1 complex subunit 2
          Gene Aliases:
          ASC1p100, p100
          Chromosome Location:
          Chr.22: 29788608 - 29838304 on Build GRCh38
          UniGene ID:
          Hs.731754
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 33848

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ASCC2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_032204.4 NP_115580.2 3, 4 392
          XM_005261775.2 XP_005261832.1 4, 5 432
          XM_011530442.2 XP_011528744.1 4, 5 432
          XM_011530443.2 XP_011528745.1 5, 6 498
          XM_011530444.2 XP_011528746.1 5, 6 545
          XM_011530445.2 XP_011528747.1 4, 5 479
          XM_011530446.2 XP_011528748.1 3, 4 362
          XM_011530450.2 XP_011528752.1 5, 6 548
          XM_011530451.2 XP_011528753.1 6, 7 622
          XM_011530452.2 XP_011528754.1 5, 6 595
          XM_011530453.2 XP_011528755.1 4, 5 450
          XM_011530454.2 XP_011528756.1 3, 4 334
          XM_011530455.2 XP_011528757.1 2, 3 263
          XM_017028991.1 XP_016884480.1 3, 4 2570
          XM_017028992.1 XP_016884481.1 3, 4 2570
          XM_017028993.1 XP_016884482.1 3, 4 2570
          XM_017028994.1 XP_016884483.1 3, 4 2570
          XM_017028996.1 XP_016884485.1 4, 5 1372
          XM_017028998.1 XP_016884487.1 4, 5 1372
          XM_017028999.1 XP_016884488.1 5, 6 498
          XM_017029000.1 XP_016884489.1 4, 5 479
          XM_017029005.1 XP_016884494.1 5, 6 548
          XM_017029006.1 XP_016884495.1 6, 7 622
          XM_017029007.1 XP_016884496.1 4, 5 450
          XM_017029008.1 XP_016884497.1 3, 4 334
          XM_017029009.1 XP_016884498.1 2, 3 263
          GenBank mRNA AK022886.1 - 5, 6 510
          AY013289.1 - 2, 3 269
          BC025368.2 - 3, 4 353 3955747

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s)

          DNA dealkylation involved in DNA repair
          transcription, DNA-templated
          regulation of transcription, DNA-templated

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0