Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ASCC2 SIRNA Assays ›
Gene Symbol:
ASCC2
Gene Name:
activating signal cointegrator 1 complex subunit 2
Gene Aliases:
ASC1p100, p100
Chromosome Location:
Chr.22: 29788608 - 29838304 on Build GRCh38
UniGene ID:
Hs.731754
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
33942
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ASCC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242906.1
NP_001229835.1
19
2540
NM_032204.4
NP_115580.2
20
2768
XM_005261775.2
XP_005261832.1
21
2808
XM_011530442.2
XP_011528744.1
21
2823
XM_011530443.2
XP_011528745.1
22
2889
XM_011530444.2
XP_011528746.1
22
2936
XM_011530445.2
XP_011528747.1
21
2870
XM_011530446.2
XP_011528748.1
20
2753
XM_011530448.2
XP_011528750.1
20
2656
XM_011530449.2
XP_011528751.1
19
2594
XM_011530450.2
XP_011528752.1
22
2939
XM_011530451.2
XP_011528753.1
23
3013
XM_011530452.2
XP_011528754.1
22
2986
XM_011530453.2
XP_011528755.1
21
2841
XM_011530454.2
XP_011528756.1
20
2725
XM_011530455.2
XP_011528757.1
19
2654
XM_017028991.1
XP_016884480.1
20
4961
XM_017028992.1
XP_016884481.1
20
4946
XM_017028993.1
XP_016884482.1
19
4886
XM_017028994.1
XP_016884483.1
19
4871
XM_017028995.1
XP_016884484.1
19
4802
XM_017028996.1
XP_016884485.1
21
3763
XM_017028997.1
XP_016884486.1
19
4787
XM_017028998.1
XP_016884487.1
21
3748
XM_017028999.1
XP_016884488.1
22
2874
XM_017029000.1
XP_016884489.1
21
2855
XM_017029001.1
XP_016884490.1
20
2641
XM_017029002.1
XP_016884491.1
19
2579
XM_017029003.1
XP_016884492.1
21
2715
XM_017029004.1
XP_016884493.1
18
2504
XM_017029005.1
XP_016884494.1
22
2924
XM_017029006.1
XP_016884495.1
23
2998
XM_017029007.1
XP_016884496.1
21
2826
XM_017029008.1
XP_016884497.1
20
2710
XM_017029009.1
XP_016884498.1
19
2639
GenBank mRNA
AK022886.1
-
22
2765
AK025241.1
-
12
1797
AK222723.1
-
12
1797
AK223033.1
-
12
1796
AL096741.1
-
7
1544
AY013289.1
-
19
2645
BC025368.2
-
20
2729
3955747
Back To Top
More Information
Gene Ontology Categories:
Function(s)
DNA dealkylation involved in DNA repair
transcription, DNA-templated
regulation of transcription, DNA-templated
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0