Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HEY2 SIRNA Assays ›
          Gene Symbol:
          HEY2
          Gene Name:
          hes related family bHLH transcription factor with YRPW motif 2
          Gene Aliases:
          CHF1, GRIDLOCK, GRL, HERP1, HESR2, HRT2, bHLHb32
          Chromosome Location:
          Chr.6: 125747592 - 125762243 on Build GRCh38
          UniGene ID:
          Hs.144287
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 3430

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HEY2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012259.2 NP_036391.1 2, 3 354
          XM_017010627.1 XP_016866116.1 2, 3 1738
          XM_017010628.1 XP_016866117.1 3, 4 1436
          XM_017010629.1 XP_016866118.1 2, 3 215
          GenBank mRNA AB044755.1 - 2, 3 259
          AF173901.1 - 2, 3 322
          AF232238.1 - 2, 3 273
          AF237949.1 - 2, 3 230
          AF311884.1 - 2, 3 163
          AJ249545.1 - 2, 3 213
          AK289943.1 - 2, 3 272
          AK303890.1 - 2, 3 176
          BC007707.2 - 2, 3 252 3945225
          BT020067.1 - 2, 3 163
          BT020068.1 - 2, 3 163

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          vasculogenesis
          muscular septum morphogenesis
          outflow tract morphogenesis
          pulmonary valve morphogenesis
          tricuspid valve morphogenesis
          tricuspid valve formation
          endocardial cushion to mesenchymal transition involved in heart valve formation
          cardiac ventricle morphogenesis
          cardiac left ventricle morphogenesis
          cardiac right ventricle morphogenesis
          ventricular trabecula myocardium morphogenesis
          transcription, DNA-templated
          Notch signaling pathway
          anterior/posterior axis specification
          positive regulation of heart rate
          negative regulation of transcription by transcription factor localization
          negative regulation of gene expression
          negative regulation of cardiac muscle cell apoptotic process
          mesenchymal cell development
          cardiac muscle hypertrophy in response to stress
          ascending aorta morphogenesis
          dorsal aorta morphogenesis
          umbilical cord morphogenesis
          cell fate commitment
          regulation of auditory receptor cell differentiation
          negative regulation of Notch signaling pathway
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          smooth muscle cell differentiation
          ventricular cardiac muscle cell development
          positive regulation of cardiac muscle cell proliferation
          cardiac epithelial to mesenchymal transition
          heart trabecula formation
          cardiac septum morphogenesis
          ventricular septum morphogenesis
          atrial septum morphogenesis
          negative regulation of transcription initiation from RNA polymerase II promoter
          labyrinthine layer blood vessel development
          arterial endothelial cell differentiation
          cardiac vascular smooth muscle cell development
          coronary vasculature morphogenesis
          pulmonary artery morphogenesis
          Notch signaling involved in heart development
          protein-DNA complex assembly
          cochlea development
          vascular smooth muscle cell development
          negative regulation of transcription regulatory region DNA binding
          negative regulation of cardiac vascular smooth muscle cell differentiation
          negative regulation of transcription from RNA polymerase II promoter involved in smooth muscle cell differentiation
          regulation of vasculogenesis
          transcription factor activity, RNA polymerase II core promoter sequence-specific
          transcription factor activity, protein binding
          RNA polymerase II activating transcription factor binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          microsatellite binding
          protein homodimerization activity
          histone deacetylase binding
          sequence-specific DNA binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0