Search
Search

RPUSD4| Gene Symbol: RPUSD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144827.1 | NP_001138299.1 | 2 | 341 | ||
| NM_032795.2 | NP_116184.2 | 2 | 341 | |||
| XM_011543039.2 | XP_011541341.1 | 2 | 311 | |||
| GenBank mRNA | AK027400.1 | - | 2 | 295 | ||
| AK225320.1 | - | 2 | 301 | |||
| AK308059.1 | - | 2 | 295 | |||
| BC014131.2 | - | 2 | 302 | 4621434 | ||
| DB051034.1 | - | 2 | 343 | |||