Search
Search

PNPT1| Gene Symbol: PNPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033109.4 | NP_149100.2 | 22 | 1844 | ||
| XM_005264629.2 | XP_005264686.1 | 22 | 1774 | |||
| XM_017005172.1 | XP_016860661.1 | 21 | 1724 | |||
| GenBank mRNA | AJ458465.1 | - | 22 | 1775 | ||
| AK307589.1 | - | 1774 | ||||
| AY027528.1 | - | 22 | 1771 | |||
| AY290863.1 | - | 4 | 234 | |||
| BC000862.2 | - | 4 | 208 | |||
| BC005986.1 | - | 4 | 197 | |||
| BC053660.1 | - | 22 | 1758 | 6062060 | ||
| CR749867.1 | - | 3 | 147 | |||