Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other N4BP2L2 SIRNA Assays ›
          Gene Symbol:
          N4BP2L2
          Gene Name:
          NEDD4 binding protein 2 like 2
          Gene Aliases:
          92M18.3, CG005, CG016, PFAAP5
          Chromosome Location:
          Chr.13: 32432488 - 32542710 on Build GRCh38
          UniGene ID:
          Hs.507680
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 35291

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: N4BP2L2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001278432.1 NP_001265361.1 7 1621
          NM_001320836.1 NP_001307765.1 7 2933
          NM_033111.4 NP_149102.3 7 1576
          XM_011534869.2 XP_011533171.1 6 2306
          XM_011534870.2 XP_011533172.1 4 1312
          XM_011534871.2 XP_011533173.1 4 1247
          XM_011534872.2 XP_011533174.1 5 1434
          XM_017020324.1 XP_016875813.1 8 2899
          XM_017020325.1 XP_016875814.1 7 2893
          XM_017020326.1 XP_016875815.1 7 2778
          XM_017020327.1 XP_016875816.1 7 2926
          XM_017020328.1 XP_016875817.1 8 2899
          XM_017020329.1 XP_016875818.1 7 2893
          XM_017020330.1 XP_016875819.1 7 2926
          XM_017020331.1 XP_016875820.1 6 2181
          XM_017020332.1 XP_016875821.1 7 2088
          XM_017020333.1 XP_016875822.1 6 2181
          XM_017020334.1 XP_016875823.1 7 2088
          XM_017020335.1 XP_016875824.1 6 2066
          XM_017020336.1 XP_016875825.1 4 1312
          XM_017020337.1 XP_016875826.1 4 1247
          XM_017020338.1 XP_016875827.1 4 1249
          XM_017020339.1 XP_016875828.1 3 1569
          XM_017020340.1 XP_016875829.1 4 1390
          XM_017020341.1 XP_016875830.1 4 1363
          XM_017020343.1 XP_016875832.1 3 1210
          XM_017020344.1 XP_016875833.1 3 1126
          XM_017020345.1 XP_016875834.1 4 1326
          XM_017020346.1 XP_016875835.1 4 1317
          GenBank mRNA AK093514.1 - 4 1317
          AK298546.1 - 6 1528
          AK310280.1 - 4 1248
          AL049802.1 - 5 1439
          U50529.1 - 5 1440

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          positive regulation of hematopoietic stem cell proliferation
          negative regulation of hematopoietic stem cell differentiation
          RNA polymerase II transcription corepressor activity
          molecular_function
          protein binding
          enzyme binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0