Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HDAC9 SIRNA Assays ›
Gene Symbol:
HDAC9
Gene Name:
histone deacetylase 9
Gene Aliases:
HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, MITR
Chromosome Location:
Chr.7: 18086942 - 19003745 on Build GRCh38
UniGene ID:
Hs.196054
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
35886
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HDAC9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204144.2
NP_001191073.1
5
582
NM_001204145.2
NP_001191074.1
3
274
NM_001204146.2
NP_001191075.1
3
443
NM_001204147.2
NP_001191076.1
2
240
NM_001204148.2
NP_001191077.1
2
240
NM_001321868.1
NP_001308797.1
4
522
NM_001321869.1
NP_001308798.1
4
522
NM_001321870.1
NP_001308799.1
4
522
NM_001321871.1
NP_001308800.1
4
522
NM_001321872.1
NP_001308801.1
4
522
NM_001321873.1
NP_001308802.1
4
522
NM_001321874.1
NP_001308803.1
3
443
NM_001321875.1
NP_001308804.1
3
443
NM_001321876.1
NP_001308805.1
3
443
NM_001321877.1
NP_001308806.1
3
443
NM_001321878.1
NP_001308807.1
3
443
NM_001321879.1
NP_001308808.1
3
443
NM_001321884.1
NP_001308813.1
3
443
NM_001321885.1
NP_001308814.1
3
443
NM_001321886.1
NP_001308815.1
4
502
NM_001321887.1
NP_001308816.1
4
333
NM_001321888.1
NP_001308817.1
3
274
NM_001321889.1
NP_001308818.1
4
502
NM_001321890.1
NP_001308819.1
3
443
NM_001321891.1
NP_001308820.1
3
443
NM_001321893.1
NP_001308822.1
4
340
NM_001321894.1
NP_001308823.1
4
340
NM_001321895.1
NP_001308824.1
3
274
NM_001321896.1
NP_001308825.1
4
612
NM_001321897.1
NP_001308826.1
4
340
NM_001321898.1
NP_001308827.1
3
274
NM_001321899.1
NP_001308828.1
4
340
NM_001321900.1
NP_001308829.1
3
443
NM_001321901.1
NP_001308830.1
3
274
NM_001321902.1
NP_001308831.1
3
274
NM_014707.3
NP_055522.1
3
274
NM_058176.2
NP_478056.1
2
129
NM_178423.2
NP_848510.1
3
274
NM_178425.3
NP_848512.1
2
129
XM_011515629.2
XP_011513931.1
3
223
XM_011515631.2
XP_011513933.1
4
522
XM_011515634.2
XP_011513936.1
3
223
XM_011515635.2
XP_011513937.1
4
1074
XM_011515636.2
XP_011513938.1
3
804
XM_011515637.2
XP_011513939.1
3
228
XM_011515638.2
XP_011513940.1
3
804
XM_011515639.2
XP_011513941.1
3
1001
XM_011515641.2
XP_011513943.1
4
1071
XM_011515642.2
XP_011513944.1
2
1800
XM_011515644.2
XP_011513946.1
4
1061
XM_011515645.2
XP_011513947.1
2
1800
XM_011515646.2
XP_011513948.1
2
1800
XM_011515648.2
XP_011513950.1
3
804
XM_011515649.2
XP_011513951.1
3
1008
XM_011515651.1
XP_011513953.1
3
204
XM_017012826.1
XP_016868315.1
3
804
XM_017012827.1
XP_016868316.1
4
863
XM_017012828.1
XP_016868317.1
3
1003
XM_017012829.1
XP_016868318.1
3
204
XM_017012830.1
XP_016868319.1
3
204
XM_017012831.1
XP_016868320.1
3
204
XM_017012832.1
XP_016868321.1
3
204
GenBank mRNA
AB018287.1
-
3
238
AJ459808.1
-
3
238
AK097178.1
-
3
223
AK296214.1
-
3
175
AK297404.1
-
3
228
AK304298.1
-
5
575
AK304343.1
-
2
160
AK304410.1
-
2
203
AK316109.1
-
3
394
AY032737.1
-
2
88
AY032738.1
-
2
88
AY197371.1
-
2
88
AY429540.1
-
3
328
BC111735.1
-
2
87
BC150328.1
-
3
249
8860495
BC152405.1
-
3
238
8862508
CR749550.1
-
2
196
DC333353.1
-
4
522
DC333611.1
-
3
443
DC369193.1
-
2
236
DC369899.1
-
2
240
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001200045, XM_008764615, XM_008764610, XM_008764613, XM_008764616, and others.
Panther Classification:
Molecular Function -
histone modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
response to amphetamine
transcription, DNA-templated
inflammatory response
heart development
chromatin modification
histone deacetylation
neuron differentiation
B cell differentiation
cellular response to insulin stimulus
peptidyl-lysine deacetylation
B cell activation
negative regulation of transcription, DNA-templated
regulation of skeletal muscle fiber development
regulation of striated muscle cell differentiation
histone H3 deacetylation
histone H4 deacetylation
positive regulation of cell migration involved in sprouting angiogenesis
transcription corepressor activity
histone deacetylase activity
protein kinase C binding
protein binding
transcription factor binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
protein deacetylase activity
histone deacetylase binding
metal ion binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0