Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LOC107984026 SIRNA Assays ›
          Gene Symbol:
          LOC107984026 AGAP11 AGAP4 AGAP5 AGAP7P AGAP9
          Gene Name:
          arf-GAP with GTPase, ANK repeat and PH domain-containing protein 4-like
          ArfGAP with GTPase domain, ankyrin repeat and PH domain 11
          ArfGAP with GTPase domain, ankyrin repeat and PH domain 4
          ArfGAP with GTPase domain, ankyrin repeat and PH domain 5
          ArfGAP with GTPase domain, ankyrin repeat and PH domain 7, pseudogene
          ArfGAP with GTPase domain, ankyrin repeat and PH domain 9
          Gene Aliases:
          -
          -
          AGAP-4, AGAP-8, AGAP8, CTGLF1, CTGLF5, MRIP2
          CTGLF2
          AGAP-7, AGAP7, CTGLF4, bA109G10.1
          AGAP-9, CTGLF6, bA301J7.2
          Chromosome Location:
          Chr.10: 46335552 - 46838268 on Build GRCh38
          Chromosome Location:
          Chr.10: 86970741 - 87010203 on Build GRCh38
          Chromosome Location:
          Chr.10: 45825592 - 45858529 on Build GRCh38
          Chromosome Location:
          Chr.10: 73674275 - 73697796 on Build GRCh38
          Chromosome Location:
          Chr.10: 46109495 - 46131678 on Build GRCh38
          Chromosome Location:
          Chr.10: 47501854 - 47523638 on Build GRCh38
          UniGene ID:
          -
          Hs.511787
          Hs.314437
          Hs.651147
          Hs.447454
          Hs.645346
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          siRNA ID 36619

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LOC107984026
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017017029.1 XP_016872518.1 10 2141
          XM_017017030.1 XP_016872519.1 9 2072
          XM_017017031.1 XP_016872520.1 9 2072
          XM_017017032.1 XP_016872521.1 11 2172
          XM_017017033.1 XP_016872522.1 12 1935
          XM_017017034.1 XP_016872523.1 11 1840
          XM_017017035.1 XP_016872524.1 7 1353
          XM_017017036.1 XP_016872525.1 5 1514
          XM_017017037.1 XP_016872526.1 7 1903
          XM_017017038.1 XP_016872527.1 5 1156
          XM_017017039.1 XP_016872528.1 7 1372
          XM_017017040.1 XP_016872529.1 7 1336
          Gene Symbol: AGAP11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_133447.1 NP_597704.1 12 2394
          GenBank mRNA AB075855.1 - 7 3798
          BC140762.1 - 8 1640 9021630
          Gene Symbol: AGAP4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001276343.2 NP_001263272.2 8 1724
          NM_001291379.1 NP_001278308.1 10 1916
          NM_133446.3 NP_597703.2 7 1655
          XM_011539269.2 XP_011537571.1 6 1262
          XM_017015664.1 XP_016871153.1 10 5586
          XM_017015665.1 XP_016871154.1 11 5454
          XM_017015666.1 XP_016871155.1 9 2489
          XM_017015667.1 XP_016871156.1 10 1759
          XM_017015668.1 XP_016871157.1 8 1509
          XM_017015669.1 XP_016871158.1 8 1417
          GenBank mRNA AF411132.1 - 7 1655
          BC048341.1 - 1 777
          BC171820.1 - 8 1709
          Gene Symbol: AGAP5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001144000.1 NP_001137472.1 8 1639
          Gene Symbol: AGAP7P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_126580.1 7 1624
          GenBank mRNA AL040331.1 - 1 366
          BY996334.1 - 1 689
          Gene Symbol: AGAP9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001190810.1 NP_001177739.1 8 1710
          GenBank mRNA AK160375.1 - 10 4626
          BC171845.1 - 8 1710

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of GTPase activity
          GTPase activator activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-4wl44:80/100.66.77.209:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0