Search
Search

ADAMTS17| Gene Symbol: ADAMTS17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139057.3 | NP_620688.2 | 4 | 878 | ||
| XM_005254872.3 | XP_005254929.1 | 4 | 1794 | |||
| XM_011521312.2 | XP_011519614.1 | 4 | 1794 | |||
| XM_017021973.1 | XP_016877462.1 | 4 | 1795 | |||
| XM_017021974.1 | XP_016877463.1 | 4 | 1797 | |||
| XM_017021975.1 | XP_016877464.1 | 4 | 1795 | |||
| XM_017021976.1 | XP_016877465.1 | 3 | 507 | |||
| XM_017021977.1 | XP_016877466.1 | 4 | 1794 | |||
| XM_017021981.1 | XP_016877470.1 | 4 | 1792 | |||
| XM_017021984.1 | XP_016877473.1 | 3 | 453 | |||
| GenBank mRNA | AJ315735.1 | - | 4 | 795 | ||
| AK131344.1 | - | 3 | 424 | |||
| DB279404.1 | - | 3 | 508 | |||