Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZFYVE27 SIRNA Assays ›
          Gene Symbol:
          ZFYVE27
          Gene Name:
          zinc finger FYVE-type containing 27
          Gene Aliases:
          PROTRUDIN, SPG33
          Chromosome Location:
          Chr.10: 97737121 - 97760907 on Build GRCh38
          UniGene ID:
          Hs.744075
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 37261

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZFYVE27
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001002261.3 NP_001002261.1 3 383
          NM_001002262.3 NP_001002262.1 4 588
          NM_001174119.1 NP_001167590.1 3 488
          NM_001174121.1 NP_001167592.1 2 315
          NM_144588.6 NP_653189.3 4 588
          XM_005269502.3 XP_005269559.1 4 577
          XM_005269503.3 XP_005269560.1 4 581
          XM_005269504.3 XP_005269561.1 4 577
          XM_005269505.4 XP_005269562.1 4 577
          XM_005269506.3 XP_005269563.1 4 577
          XM_005269508.3 XP_005269565.1 3 506
          XM_005269509.3 XP_005269566.1 3 510
          XM_011539252.2 XP_011537554.1 4 577
          XM_011539253.2 XP_011537555.1 4 577
          XM_011539254.2 XP_011537556.1 4 552
          XM_011539255.2 XP_011537557.1 4 559
          XM_017015644.1 XP_016871133.1 4 577
          XM_017015645.1 XP_016871134.1 4 577
          XM_017015646.1 XP_016871135.1 4 577
          XM_017015647.1 XP_016871136.1 4 581
          XM_017015650.1 XP_016871139.1 4 552
          XM_017015651.1 XP_016871140.1 3 506
          XM_017015652.1 XP_016871141.1 3 510
          XM_017015653.1 XP_016871142.1 3 506
          XM_017015654.1 XP_016871143.1 3 510
          GenBank mRNA AK057481.1 - 4 559
          AK074876.1 - 4 569
          AK296295.1 - 2 315
          AK296588.1 - 3 433
          AK300438.1 - 4 581
          AL561314.3 - 4 500
          AL834235.1 - 3 317
          BC030621.2 - 4 581 4818199
          BF508037.1 - 4 551
          BI758753.1 - 4 576
          BX647836.1 - 4 523
          KU178847.1 - 2 178
          KU178848.1 - 2 178

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane traffic protein membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          neuron projection development
          neurotrophin TRK receptor signaling pathway
          protein localization to plasma membrane
          protein binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0