Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TSNARE1 SIRNA Assays ›
Gene Symbol:
TSNARE1
Gene Name:
t-SNARE domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.8: 142212080 - 142403291 on Build GRCh38
UniGene ID:
Hs.370931
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
37692
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TSNARE1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001291931.1
NP_001278860.1
8
709
NM_145003.4
NP_659440.2
10
1366
XM_011516912.1
XP_011515214.1
9
1220
XM_011516913.2
XP_011515215.1
10
1408
XM_011516914.1
XP_011515216.1
10
1322
XM_011516915.2
XP_011515217.1
11
1408
XM_011516916.1
XP_011515218.1
10
1411
XM_011516917.2
XP_011515219.1
10
1248
XM_011516918.1
XP_011515220.1
8
1073
XM_011516919.1
XP_011515221.1
9
1223
XM_011516920.1
XP_011515222.1
7
566
XM_011516921.1
XP_011515223.1
9
1220
XM_011516922.1
XP_011515224.1
9
1223
XM_011516923.2
XP_011515225.1
5
1550
XM_017013175.1
XP_016868664.1
9
1223
XM_017013176.1
XP_016868665.1
9
1223
XM_017013177.1
XP_016868666.1
9
1223
XM_017013178.1
XP_016868667.1
9
1217
XM_017013179.1
XP_016868668.1
9
1223
XM_017013180.1
XP_016868669.1
9
1220
GenBank mRNA
AK055726.1
-
10
1295
BC065226.1
-
8
628
BC113556.1
-
9
1220
8322608
BC126343.1
-
9
1223
8992059
BC143684.1
-
9
1217
9052198
BC143686.1
-
9
1223
9052200
Back To Top
More Information
Panther Classification:
Molecular Function -
SNARE protein
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
vesicle fusion
vesicle docking
SNARE binding
SNAP receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0