Search
Search

ZNF23| Gene Symbol: ZNF23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304492.1 | NP_001291421.1 | 5 | 487 | ||
| NM_001304493.1 | NP_001291422.1 | 7 | 612 | |||
| NM_001304494.1 | NP_001291423.1 | 3 | 205 | |||
| NM_145911.2 | NP_666016.1 | 6 | 1059 | |||
| GenBank mRNA | AF507946.1 | - | 6 | 1050 | ||
| AK091039.1 | - | 5 | 482 | |||
| AK297130.1 | - | 3 | 205 | |||
| AL080123.1 | - | 1 | 89 | |||
| AL833815.1 | - | 2 | 829 | |||
| BC007256.1 | - | 6 | 1050 | 3028040 | ||
| BC048974.1 | - | 7 | 606 | 4816965 | ||
| BT007400.1 | - | 3 | 236 | |||
| BX537507.1 | - | 1 | 2861 | |||