Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other USP54 SIRNA Assays ›
Gene Symbol:
USP54
Gene Name:
ubiquitin specific peptidase 54
Gene Aliases:
C10orf29, bA137L10.3, bA137L10.4
Chromosome Location:
Chr.10: 73497538 - 73626844 on Build GRCh38
UniGene ID:
Hs.657355
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
39267
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: USP54
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320437.1
NP_001307366.1
18
3264
NM_152586.3
NP_689799.3
18
3362
NR_135249.1
19
3749
NR_135250.1
14
2744
XM_005269582.2
XP_005269639.1
17
6440
XM_011539368.2
XP_011537670.1
18
6506
XM_017015759.1
XP_016871248.1
20
5935
XM_017015760.1
XP_016871249.1
20
4837
XM_017015761.1
XP_016871250.1
20
5675
XM_017015762.1
XP_016871251.1
20
4845
XM_017015763.1
XP_016871252.1
20
4139
XM_017015764.1
XP_016871253.1
20
4202
XM_017015765.1
XP_016871254.1
21
4455
XM_017015766.1
XP_016871255.1
20
6061
XM_017015767.1
XP_016871256.1
20
5720
XM_017015768.1
XP_016871257.1
20
3891
XM_017015769.1
XP_016871258.1
20
3499
XM_017015770.1
XP_016871259.1
20
4160
XM_017015771.1
XP_016871260.1
19
6677
XM_017015772.1
XP_016871261.1
19
6683
XM_017015773.1
XP_016871262.1
18
6617
XM_017015774.1
XP_016871263.1
19
5603
XM_017015775.1
XP_016871264.1
20
5481
XM_017015776.1
XP_016871265.1
19
5986
XM_017015777.1
XP_016871266.1
19
5645
XM_017015778.1
XP_016871267.1
18
6563
XM_017015779.1
XP_016871268.1
19
6683
XM_017015780.1
XP_016871269.1
19
6683
XM_017015781.1
XP_016871270.1
18
6512
XM_017015782.1
XP_016871271.1
19
4263
XM_017015783.1
XP_016871272.1
19
5310
XM_017015784.1
XP_016871273.1
18
3256
XM_017015785.1
XP_016871274.1
18
6563
XM_017015786.1
XP_016871275.1
18
3753
GenBank mRNA
AK094637.1
-
1
456
AK126497.1
-
6
1717
AK127213.1
-
5
1297
AK127267.1
-
14
2744
AK303633.1
-
3
955
AL832102.1
-
1
402
BC026037.1
-
1
210
BC064967.1
-
1
398
BC130633.1
-
3
946
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
Gene Ontology Categories:
Function(s)
Process(es)
protein deubiquitination
protein binding
thiol-dependent ubiquitinyl hydrolase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0