Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRIMPOL SIRNA Assays ›
Gene Symbol:
PRIMPOL
Gene Name:
primase and DNA directed polymerase
Gene Aliases:
CCDC111, MYP22
Chromosome Location:
Chr.4: 184649613 - 184694959 on Build GRCh38
UniGene ID:
Hs.481307
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
39630
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRIMPOL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001300767.1
NP_001287696.1
7
1117
NM_001300768.1
NP_001287697.1
8
1356
NM_152683.3
NP_689896.1
8
1356
XM_011531719.1
XP_011530021.1
8
1233
XM_011531720.1
XP_011530022.1
8
1271
XM_011531721.2
XP_011530023.1
8
1233
XM_011531723.1
XP_011530025.1
7
1058
XM_011531724.2
XP_011530026.1
6
1015
XM_011531725.1
XP_011530027.1
6
929
XM_011531726.2
XP_011530028.1
5
776
XM_011531729.1
XP_011530031.1
7
1062
XM_011531730.2
XP_011530032.1
7
1077
XM_017007864.1
XP_016863353.1
8
1271
XM_017007865.1
XP_016863354.1
8
1271
XM_017007866.1
XP_016863355.1
8
1271
XM_017007867.1
XP_016863356.1
8
1233
XM_017007868.1
XP_016863357.1
8
1233
XM_017007871.1
XP_016863360.1
6
1014
XM_017007872.1
XP_016863361.1
6
924
XM_017007873.1
XP_016863362.1
7
1103
XM_017007874.1
XP_016863363.1
6
922
XM_017007875.1
XP_016863364.1
7
1062
GenBank mRNA
AK057729.1
-
8
1139
AK307317.1
-
7
1062
BC047467.1
-
7
1063
BC064600.1
-
8
1197
5786393
BX647575.1
-
8
1244
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA metabolism protein
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
mitochondrial DNA replication
DNA replication, synthesis of RNA primer
response to UV
translesion synthesis
replication fork processing
chromatin binding
DNA-directed DNA polymerase activity
DNA primase activity
protein binding
manganese ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0