Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HIF3A SIRNA Assays ›
Gene Symbol:
HIF3A
Gene Name:
hypoxia inducible factor 3 alpha subunit
Gene Aliases:
HIF-3A, HIF3-alpha-1, IPAS, MOP7, PASD7, bHLHe17
Chromosome Location:
Chr.19: 46297046 - 46343433 on Build GRCh38
UniGene ID:
Hs.420830
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
39813
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HIF3A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022462.4
NP_071907.4
7
869
NM_152794.3
NP_690007.1
8
965
NM_152795.3
NP_690008.2
8
933
NM_152796.2
NP_690009.1
7
869
XM_005259152.4
XP_005259209.1
9
1061
XM_005259153.3
XP_005259210.2
6
732
XM_005259155.4
XP_005259212.1
9
1061
XM_005259156.4
XP_005259213.1
9
1061
XM_017027132.1
XP_016882621.1
9
1061
XM_017027133.1
XP_016882622.1
9
1061
XM_017027134.1
XP_016882623.1
8
931
XM_017027135.1
XP_016882624.1
8
965
XM_017027136.1
XP_016882625.1
9
1061
XM_017027137.1
XP_016882626.1
8
965
XM_017027138.1
XP_016882627.1
8
931
XM_017027139.1
XP_016882628.1
7
855
XM_017027140.1
XP_016882629.1
7
785
XM_017027141.1
XP_016882630.1
7
827
XM_017027142.1
XP_016882631.1
7
855
GenBank mRNA
AB054067.1
-
8
896
AB295039.1
-
8
914
AF463492.1
-
8
928
AK021421.1
-
7
818
AK021653.1
-
8
959
AK024095.1
-
6
880
AK027725.1
-
8
914
AK297828.1
-
7
855
AK299699.1
-
6
779
AK304747.1
-
8
997
AK314680.1
-
8
965
BC026308.1
-
7
1162
4811824
BC080551.1
-
8
904
6250259
BE250295.1
-
2
116
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_022528, AJ277827.
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
transcription from RNA polymerase II promoter
apoptotic process
positive regulation of transcription from RNA polymerase II promoter
regulation of transcription from RNA polymerase II promoter in response to hypoxia
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
protein dimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0