Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BDNF SIRNA Assays ›
Gene Symbol:
BDNF
Gene Name:
brain derived neurotrophic factor
Gene Aliases:
ANON2, BULN2
Chromosome Location:
Chr.11: 27654893 - 27722058 on Build GRCh38
UniGene ID:
Hs.502182
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
40329
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BDNF
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001143805.1
NP_001137277.1
2
315
NM_001143806.1
NP_001137278.1
2
530
NM_001143807.1
NP_001137279.1
2
416
NM_001143808.1
NP_001137280.1
2
252
NM_001143809.1
NP_001137281.1
2
262
NM_001143810.1
NP_001137282.1
3
545
NM_001143811.1
NP_001137283.1
4
662
NM_001143812.1
NP_001137284.1
2
404
NM_001143813.1
NP_001137285.1
2
486
NM_001143814.1
NP_001137286.1
3
611
NM_001143816.1
NP_001137288.1
2
1009
NM_001709.4
NP_001700.2
2
504
NM_170731.4
NP_733927.1
2
826
NM_170732.4
NP_733928.1
2
613
NM_170733.3
NP_733929.1
2
516
NM_170734.3
NP_733930.1
2
464
NM_170735.5
NP_733931.1
1
1243
XM_011520280.2
XP_011518582.1
2
1410
GenBank mRNA
AF400438.1
-
1
176
AK289763.1
-
2
312
AK289853.1
-
2
213
AY054392.2
-
2
517
AY054393.2
-
2
259
AY054394.2
-
2
474
AY054395.2
-
2
557
AY054396.2
-
2
315
AY054397.2
-
2
398
AY054398.2
-
2
445
AY054399.2
-
3
330
AY054400.2
-
2
431
AY054406.3
-
2
299
AY656701.1
-
1
158
BC029795.1
-
2
489
5193877
CX751420.1
-
2
383
EF674517.1
-
2
315
EF674518.1
-
2
530
EF674519.1
-
2
613
EF674520.1
-
2
416
EF674521.1
-
2
516
EF689009.1
-
1
1257
EF689010.1
-
2
1023
EF689011.1
-
2
252
EF689012.1
-
2
262
EF689013.1
-
2
404
EF689014.1
-
2
486
EF689015.1
-
2
504
EF689016.1
-
3
611
EF689017.1
-
3
363
EF689018.1
-
2
464
EF689019.1
-
3
545
EF689020.1
-
4
662
EF689021.1
-
2
826
KP229412.1
-
1
158
KU177943.1
-
1
158
KU177944.1
-
1
158
KU177945.1
-
1
158
M61176.1
-
1
720
X60201.1
-
2
454
X91251.1
-
2
442
Back To Top
More Information
Panther Classification:
Molecular Function -
neurotrophic factor
Gene Ontology Categories:
Function(s)
Process(es)
cell-cell signaling
nervous system development
axon guidance
synapse assembly
brain-derived neurotrophic factor receptor signaling pathway
positive regulation of brain-derived neurotrophic factor receptor signaling pathway
negative regulation of neuron apoptotic process
collateral sprouting
positive regulation of collateral sprouting
positive regulation of synapse assembly
neurotrophin TRKB receptor binding
growth factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0