Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NFATC3 SIRNA Assays ›
          Gene Symbol:
          NFATC3
          Gene Name:
          nuclear factor of activated T-cells 3
          Gene Aliases:
          NFAT4, NFATX
          Chromosome Location:
          Chr.16: 68085366 - 68229259 on Build GRCh38
          UniGene ID:
          Hs.436585
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 40755

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NFATC3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004555.3 NP_004546.1 2 1346
          NM_173163.2 NP_775186.1 2 1346
          NM_173165.2 NP_775188.1 2 1346
          GenBank mRNA AK290370.1 - 2 1086
          BC001050.2 - 2 1064 3505967
          EU887605.1 - 2 1030
          EU887606.1 - 2 1030
          EU887607.1 - 2 1030
          EU887608.1 - 2 1357
          EU887609.1 - 2 1357
          EU887610.1 - 2 1357
          EU887611.1 - 2 1075
          EU887612.1 - 2 1075
          EU887613.1 - 2 1075
          EU887614.1 - 2 983
          EU887615.1 - 2 983
          EU887616.1 - 2 983
          EU887617.1 - 2 988
          EU887618.1 - 2 988
          EU887619.1 - 2 988
          EU887620.1 - 2 1025
          EU887621.1 - 2 1025
          EU887622.1 - 2 1025
          EU887623.1 - 2 1429
          EU887624.1 - 2 1429
          EU887625.1 - 2 1429
          L41067.1 - 2 1240
          U14510.1 - 2 1054
          U85428.1 - 2 1054
          U85429.1 - 2 1054
          U85430.1 - 2 1054

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          Rel homology transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          inflammatory response
          Fc-epsilon receptor signaling pathway
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of pri-miRNA transcription from RNA polymerase II promoter
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-kjp6x:80/100.66.78.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0