Search
Search

DNAH17| Gene Symbol: DNAH17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173628.3 | NP_775899.3 | 7 | 1131 | ||
| XM_011525416.2 | XP_011523718.1 | 7 | 1167 | |||
| GenBank mRNA | AK097776.1 | - | 3 | 218 | ||
| BC105107.1 | - | 2 | 175 | |||
| BC112284.1 | - | 2 | 175 | |||