Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PIP5K1A SIRNA Assays ›
Gene Symbol:
PIP5K1A
PIPSL
Gene Name:
phosphatidylinositol-4-phosphate 5-kinase type 1 alpha
PIP5K1A and PSMD4-like, pseudogene
Gene Aliases:
-
PIP5K1L1, PIP5K1P3, PSMD4P2, bA429H9.1
Chromosome Location:
Chr.1: 151198003 - 151249536 on Build GRCh38
Chromosome Location:
Chr.10: 93958140 - 93961915 on Build GRCh38
UniGene ID:
Hs.655131
Hs.149255
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
414
Purification
In-Vivo Ready
Standard
HPLC
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
6,301.00
Your Price
Online offer:
6301.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PIP5K1A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135637.1
NP_001129109.1
9, 10
1650
NM_001135638.1
NP_001129110.1
10, 11
1689
NM_003557.2
NP_003548.1
9, 10
1650
XM_005245525.4
XP_005245582.1
11, 12
1930
XM_005245527.3
XP_005245584.1
10, 11
1679
XM_005245528.4
XP_005245585.1
10, 11
1894
XM_005245530.4
XP_005245587.1
10, 11
1252
XM_005245531.3
XP_005245588.1
9, 10
1646
XM_005245532.3
XP_005245589.1
9, 10
1579
XM_006711563.3
XP_006711626.1
11, 12
1936
XM_006711564.3
XP_006711627.1
10, 11
1648
XM_006711567.3
XP_006711630.1
11, 12
1236
XM_006711568.3
XP_006711631.2
10, 11
1267
XM_011510043.2
XP_011508345.1
9, 10
1231
XM_011510045.2
XP_011508347.1
10, 11
1249
XM_011510046.2
XP_011508348.1
9, 10
1724
XM_011510048.2
XP_011508350.1
11, 12
1246
XM_011510049.2
XP_011508351.1
11, 12
1233
XM_017002439.1
XP_016857928.1
9, 10
1646
XM_017002440.1
XP_016857929.1
9, 10
1643
XM_017002441.1
XP_016857930.1
11, 12
1252
GenBank mRNA
AK291015.1
-
9, 10
1593
AK302268.1
-
3, 4
365
BC007833.2
-
9, 10
1642
4098514
U78575.1
-
9, 10
1598
U78576.1
-
10, 11
1658
U78577.1
-
9, 10
1599
U78578.1
-
2, 3
174
Gene Symbol: PIPSL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_002319.2
1
1597
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001042621, XM_008761315, XM_008761316, AY724476.
This siRNA also targets the following transcripts in Mouse: NM_008847, NM_001293707, AK098097, AK150898, BC031774, and others.
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
glycerophospholipid metabolic process
phosphatidylinositol biosynthetic process
phagocytosis
signal transduction
phospholipid biosynthetic process
fibroblast migration
regulation of phosphatidylinositol 3-kinase signaling
cell migration
keratinocyte differentiation
actin cytoskeleton reorganization
phosphatidylinositol phosphorylation
focal adhesion assembly
cell chemotaxis
protein targeting to plasma membrane
activation of GTPase activity
ruffle assembly
1-phosphatidylinositol-3-phosphate 5-kinase activity
protein binding
ATP binding
1-phosphatidylinositol-4-phosphate 5-kinase activity
kinase binding
1-phosphatidylinositol-5-kinase activity
1-phosphatidylinositol-3-phosphate 4-kinase activity
phosphatidylinositol-3,4-bisphosphate 5-kinase activity
phosphatidylinositol phosphate kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0