Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BAD SIRNA Assays ›
          Gene Symbol:
          BAD
          Gene Name:
          BCL2 associated agonist of cell death
          Gene Aliases:
          BBC2, BCL2L8
          Chromosome Location:
          Chr.11: 64269828 - 64284704 on Build GRCh38
          UniGene ID:
          Hs.370254
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 41903

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BAD
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004322.3 NP_004313.1 2 691
          NM_032989.2 NP_116784.1 3 437
          GenBank mRNA AB451254.1 - 2 355
          AB451378.1 - 2 355
          AF021792.1 - 2 355
          AF031523.1 - 2 355
          AK291863.1 - 3 437
          AK303051.1 - 3 437
          BC001901.2 - 2 363 3537914
          BC095431.1 - 3 402 30377088
          BG435829.1 - 2 621
          BG748336.1 - 3 425
          BT006678.1 - 2 355
          CR541935.1 - 2 355
          CR541959.1 - 2 355
          U66879.1 - 2 421

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          release of cytochrome c from mitochondria
          protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
          glucose catabolic process
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          spermatogenesis
          extrinsic apoptotic signaling pathway via death domain receptors
          intrinsic apoptotic signaling pathway in response to DNA damage
          response to glucose
          positive regulation of autophagy
          positive regulation of mitochondrial membrane potential
          suppression by virus of host apoptotic process
          cytokine-mediated signaling pathway
          cerebral cortex development
          positive regulation of insulin secretion
          response to estradiol
          response to progesterone
          positive regulation of glucokinase activity
          response to testosterone
          response to oleic acid
          positive regulation of insulin secretion involved in cellular response to glucose stimulus
          response to drug
          response to hydrogen peroxide
          glucose homeostasis
          positive regulation of apoptotic process
          response to amino acid
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          type B pancreatic cell proliferation
          response to ethanol
          positive regulation of B cell differentiation
          positive regulation of T cell differentiation
          positive regulation of proteolysis
          negative regulation of cytolysis
          ADP metabolic process
          ATP metabolic process
          regulation of mitochondrial membrane permeability
          pore complex assembly
          positive regulation of epithelial cell proliferation
          response to glucocorticoid
          response to calcium ion
          positive regulation of apoptotic process by virus
          cellular process regulating host cell cycle in response to virus
          cellular response to chromate
          cellular response to mechanical stimulus
          cellular response to nicotine
          cellular response to lipid
          cellular response to hypoxia
          positive regulation of release of cytochrome c from mitochondria
          extrinsic apoptotic signaling pathway
          extrinsic apoptotic signaling pathway in absence of ligand
          intrinsic apoptotic signaling pathway
          activation of cysteine-type endopeptidase activity
          positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
          positive regulation of neuron death
          positive regulation of type B pancreatic cell development
          positive regulation of intrinsic apoptotic signaling pathway
          protein binding
          phospholipid binding
          lipid binding
          cysteine-type endopeptidase activator activity involved in apoptotic process
          protein kinase binding
          protein phosphatase 2B binding
          protein kinase B binding
          protein heterodimerization activity
          14-3-3 protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0