Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAHD2A SIRNA Assays ›
Gene Symbol:
FAHD2A
Gene Name:
fumarylacetoacetate hydrolase domain containing 2A
Gene Aliases:
CGI-105
Chromosome Location:
Chr.2: 95402691 - 95416702 on Build GRCh38
UniGene ID:
Hs.531247
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
42324
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAHD2A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_016044.2
NP_057128.2
3
491
XM_011511282.2
XP_011509584.1
3
1240
XM_011511283.2
XP_011509585.1
4
617
XM_011511284.2
XP_011509586.1
4
746
XM_011511285.2
XP_011509587.1
4
614
XM_011511286.2
XP_011509588.1
3
500
XM_011511287.2
XP_011509589.1
3
611
XM_011511288.2
XP_011509590.1
3
1240
XM_011511289.2
XP_011509591.1
3
1240
XM_011511290.2
XP_011509592.1
3
1240
XM_017004236.1
XP_016859725.1
4
617
XM_017004237.1
XP_016859726.1
4
746
XM_017004238.1
XP_016859727.1
4
614
XM_017004239.1
XP_016859728.1
3
500
XM_017004240.1
XP_016859729.1
3
611
XM_017004241.1
XP_016859730.1
3
1240
XM_017004242.1
XP_016859731.1
3
500
XM_017004243.1
XP_016859732.1
3
1240
XM_017004244.1
XP_016859733.1
3
500
XM_017004245.1
XP_016859734.1
3
1240
XM_017004246.1
XP_016859735.1
3
1240
XM_017004247.1
XP_016859736.1
3
500
XM_017004248.1
XP_016859737.1
3
611
XM_017004249.1
XP_016859738.1
3
491
XM_017004250.1
XP_016859739.1
3
611
GenBank mRNA
AF151863.1
-
3
489
BC009403.2
-
3
465
4309334
BC110911.1
-
3
463
6064857
Back To Top
More Information
Panther Classification:
Molecular Function -
hydrolase
Gene Ontology Categories:
Function(s)
Process(es)
metabolic process
hydrolase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0