Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CASP1 SIRNA Assays ›
          Gene Symbol:
          CASP1
          Gene Name:
          caspase 1
          Gene Aliases:
          ICE, IL1BC, P45
          Chromosome Location:
          Chr.11: 105025508 - 105035591 on Build GRCh38
          UniGene ID:
          Hs.2490
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 42626

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CASP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001223.4 NP_001214.1 7 1044
          NM_001257118.2 NP_001244047.1 8 1107
          NM_001257119.2 NP_001244048.1 7 1044
          NM_033292.3 NP_150634.1 8 1107
          NM_033293.3 NP_150635.1 7 828
          NM_033294.3 NP_150636.1 6 684
          NM_033295.3 NP_150637.1 3 159
          XM_017018393.1 XP_016873882.1 8 1455
          XM_017018394.1 XP_016873883.1 8 1455
          XM_017018395.1 XP_016873884.1 7 1392
          XM_017018396.1 XP_016873885.1 8 1364
          GenBank mRNA AK223503.1 - 8 1099
          AK290114.1 - 8 1098
          AK290122.1 - 7 1035
          AK301037.1 - 8 1049
          AK310059.1 - 6 918
          AK313516.1 - 8 1093
          AY660536.1 - 8 1001
          BC041689.2 - 6 993
          BC062327.1 - 7 794 5477096
          M87507.1 - 8 1080
          U13697.1 - 6 1000
          U13698.1 - 6 784
          U13699.1 - 5 640
          U13700.1 - 2 115
          X65019.1 - 7 1070

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          proteolysis
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          signal transduction
          regulation of autophagy
          protein processing
          response to lipopolysaccharide
          interleukin-1 beta production
          response to ATP
          regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of interleukin-1 alpha secretion
          positive regulation of interleukin-1 beta secretion
          regulation of inflammatory response
          mitochondrial depolarization
          membrane hyperpolarization
          pyroptosis
          cellular response to mechanical stimulus
          cellular response to organic substance
          programmed necrotic cell death
          toxin transport
          endopeptidase activity
          cysteine-type endopeptidase activity
          protein binding
          cysteine-type endopeptidase activator activity involved in apoptotic process
          cysteine-type endopeptidase activity involved in apoptotic process

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0