Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CHEK1 SIRNA Assays ›
          Gene Symbol:
          CHEK1
          Gene Name:
          checkpoint kinase 1
          Gene Aliases:
          CHK1
          Chromosome Location:
          Chr.11: 125624910 - 125676259 on Build GRCh38
          UniGene ID:
          Hs.24529
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 42823

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CHEK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001114121.2 NP_001107593.1 4, 5 1254
          NM_001114122.2 NP_001107594.1 4, 5 1254
          NM_001244846.1 NP_001231775.1 4, 5 1254
          NM_001274.5 NP_001265.2 4, 5 597
          NR_045204.1 3, 4 1030
          NR_045205.1 4, 5 791
          XM_011542560.2 XP_011540862.1 3, 4 1256
          XM_011542561.2 XP_011540863.1 3, 4 1256
          XM_011542562.2 XP_011540864.1 3, 4 506
          XM_011542563.2 XP_011540865.1 3, 4 571
          XM_017017146.1 XP_016872635.1 4, 5 441
          GenBank mRNA AB451222.1 - 3, 4 357
          AB451345.1 - 3, 4 357
          AF016582.1 - 3, 4 391
          AF032874.1 - 3, 4 357
          AK292549.1 - 4, 5 791
          AK293143.1 - 4, 5 419
          AK299783.1 - 3, 4 191
          AK300082.1 - 3, 4 440
          BC004202.2 - 4, 5 416 3530606
          BC017575.1 - 4, 5 515 4091997
          BG468734.1 - 3, 4 456
          DA567616.1 - 4, 5 488
          JF289264.1 - 3, 4 232

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA damage checkpoint
          G2/M transition of mitotic cell cycle
          DNA replication
          DNA repair
          cellular response to DNA damage stimulus
          DNA damage induced protein phosphorylation
          regulation of double-strand break repair via homologous recombination
          regulation of transcription from RNA polymerase II promoter in response to UV-induced DNA damage
          peptidyl-threonine phosphorylation
          G2 DNA damage checkpoint
          histone H3-T11 phosphorylation
          regulation of cell proliferation
          negative regulation of mitotic nuclear division
          regulation of mitotic centrosome separation
          chromatin-mediated maintenance of transcription
          cellular response to mechanical stimulus
          replicative senescence
          regulation of signal transduction by p53 class mediator
          regulation of histone H3-K9 acetylation
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          kinase activity
          histone kinase activity (H3-T11 specific)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0