Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FGF2 SIRNA Assays ›
          Gene Symbol:
          FGF2
          Gene Name:
          fibroblast growth factor 2
          Gene Aliases:
          BFGF, FGF-2, FGFB, HBGF-2
          Chromosome Location:
          Chr.4: 122826708 - 122898235 on Build GRCh38
          UniGene ID:
          Hs.284244
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 4286

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FGF2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002006.4 NP_001997.5 2 701
          GenBank mRNA AB451321.1 - 2 234
          AB451450.1 - 2 234
          AY820133.1 - 2 207
          BC111737.1 - 3 108
          J04513.1 - 2 700
          M17599.1 - 2 240
          M27968.1 - 2 573
          S47380.1 - 2 119

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          growth factor intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPK activity
          branching involved in ureteric bud morphogenesis
          positive regulation of endothelial cell proliferation
          cell migration involved in sprouting angiogenesis
          phosphatidylinositol biosynthetic process
          chemotaxis
          signal transduction
          Ras protein signal transduction
          nervous system development
          positive regulation of cell proliferation
          fibroblast growth factor receptor signaling pathway
          organ morphogenesis
          negative regulation of fibroblast migration
          positive regulation of phospholipase C activity
          regulation of phosphatidylinositol 3-kinase signaling
          growth factor dependent regulation of skeletal muscle satellite cell proliferation
          peptidyl-tyrosine phosphorylation
          extracellular matrix organization
          hyaluronan catabolic process
          inositol phosphate biosynthetic process
          somatic stem cell population maintenance
          phosphatidylinositol-3-phosphate biosynthetic process
          wound healing
          positive regulation of cell fate specification
          positive regulation of MAP kinase activity
          positive regulation of blood vessel endothelial cell migration
          negative regulation of blood vessel endothelial cell migration
          positive regulation of GTPase activity
          positive regulation of phosphatidylinositol 3-kinase activity
          regulation of angiogenesis
          positive regulation of angiogenesis
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          embryonic morphogenesis
          positive chemotaxis
          release of sequestered calcium ion into cytosol
          positive regulation of cell division
          positive regulation of cardiac muscle cell proliferation
          negative regulation of cell death
          chondroblast differentiation
          negative regulation of wound healing
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of sprouting angiogenesis
          regulation of endothelial cell chemotaxis to fibroblast growth factor
          positive regulation of endothelial cell chemotaxis to fibroblast growth factor
          protein tyrosine kinase activity
          Ras guanyl-nucleotide exchange factor activity
          fibroblast growth factor receptor binding
          cytokine activity
          protein binding
          growth factor activity
          heparin binding
          1-phosphatidylinositol-3-kinase activity
          ligand-dependent nuclear receptor transcription coactivator activity
          chemoattractant activity
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0