Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IRF5 SIRNA Assays ›
Gene Symbol:
IRF5
Gene Name:
interferon regulatory factor 5
Gene Aliases:
SLEB10
Chromosome Location:
Chr.7: 128937737 - 128950042 on Build GRCh38
UniGene ID:
Hs.521181
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
4307
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IRF5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098627.3
NP_001092097.2
3
432
NM_001098629.2
NP_001092099.1
3
339
NM_001098630.2
NP_001092100.1
3
339
NM_001242452.2
NP_001229381.1
3
339
NM_032643.4
NP_116032.1
3
255
XM_005250317.3
XP_005250374.1
3
486
XM_006715974.2
XP_006716037.1
3
422
XM_011516158.2
XP_011514460.1
3
544
XM_011516159.2
XP_011514461.1
3
453
XM_011516160.1
XP_011514462.1
3
324
XM_011516161.1
XP_011514463.1
3
354
XM_011516162.1
XP_011514464.1
3
354
XM_011516163.2
XP_011514465.1
3
540
XM_011516164.1
XP_011514466.1
3
422
GenBank mRNA
AK293654.1
-
3
339
AK297083.1
-
3
298
AY504946.1
-
3
264
AY504947.1
-
3
287
AY693665.1
-
3
336
AY693666.1
-
3
266
AY693668.1
-
3
336
AY693669.1
-
3
255
BC004139.2
-
3
255
2963923
BC004201.2
-
3
255
2963923
BU540632.1
-
2
101
BU950658.1
-
3
338
DC427600.1
-
3
432
DQ277633.1
-
3
255
DQ277634.1
-
3
264
EU258895.1
-
2
92
EU258896.1
-
3
247
EU258897.1
-
3
346
EU258898.1
-
3
430
JQ950681.1
-
2
107
JQ950683.1
-
1
34
JQ950684.1
-
1
31
KP843527.1
-
3
315
KP843528.1
-
3
315
U51127.1
-
3
318
Back To Top
More Information
Panther Classification:
Molecular Function -
winged helix/forkhead transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
response to peptidoglycan
response to muramyl dipeptide
positive regulation of interferon-alpha production
positive regulation of interferon-beta production
positive regulation of interleukin-12 production
positive regulation of apoptotic process
positive regulation of transcription from RNA polymerase II promoter
defense response to virus
interferon-gamma-mediated signaling pathway
type I interferon signaling pathway
regulatory region DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0