Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SEMA4F SIRNA Assays ›
          Gene Symbol:
          SEMA4F
          Gene Name:
          ssemaphorin 4F
          Gene Aliases:
          M-SEMA, PRO2353, S4F, SEMAM, SEMAW, m-Sema-M
          Chromosome Location:
          Chr.2: 74654228 - 74699013 on Build GRCh38
          UniGene ID:
          Hs.25887
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 43351

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SEMA4F
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001271661.1 NP_001258590.1 1 282
          NM_001271662.1 NP_001258591.1 1 282
          NM_004263.4 NP_004254.2 1 282
          NR_073398.1 1 282
          NR_073399.1 1 282
          XM_011532475.2 XP_011530777.1 1 265
          XM_011532476.2 XP_011530778.1 1 265
          XM_011532477.2 XP_011530779.1 1 265
          XM_017003172.1 XP_016858661.1 1 255
          XM_017003173.1 XP_016858662.1 1 60
          XM_017003174.1 XP_016858663.1 1 281
          GenBank mRNA AB021292.1 - 1 148
          AB022317.1 - 1 148
          AK075384.1 - 1 235
          AK291023.1 - 1 230
          AK304358.1 - 1 233
          AK308166.1 - 1 157
          AK312728.1 - 1 258
          AL136552.1 - 1 244
          BC018361.1 - 1 221 4215248
          BC038411.1 - 1 183 5179587
          DC306380.1 - 1 282
          KU178531.1 - 1 133
          KU178532.1 - 1 133
          KU178533.1 - 1 133

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane-bound signaling molecule intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          neural crest cell migration
          cell-cell signaling
          nervous system development
          axon guidance
          positive regulation of cell migration
          retinal ganglion cell axon guidance
          negative regulation of axon extension involved in axon guidance
          negative chemotaxis
          semaphorin-plexin signaling pathway
          semaphorin receptor binding
          neuropilin binding
          chemorepellent activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0