Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CNOT2 SIRNA Assays ›
          Gene Symbol:
          CNOT2
          Gene Name:
          CCR4-NOT transcription complex subunit 2
          Gene Aliases:
          CDC36, HSPC131, NOT2, NOT2H
          Chromosome Location:
          Chr.12: 70242993 - 70354993 on Build GRCh38
          UniGene ID:
          Hs.133350
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 43390

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CNOT2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001199302.1 NP_001186231.1 4 536
          NM_001199303.1 NP_001186232.1 3 789
          NM_014515.5 NP_055330.1 3 667
          NR_037615.1 4 1013
          XM_006719429.3 XP_006719492.1 4 892
          XM_006719430.3 XP_006719493.1 4 726
          XM_006719432.3 XP_006719495.1 3 668
          XM_006719433.3 XP_006719496.1 3 668
          XM_011538399.1 XP_011536701.1 3 622
          XM_011538400.2 XP_011536702.1 4 855
          XM_017019348.1 XP_016874837.1 5 913
          XM_017019349.1 XP_016874838.1 5 1079
          XM_017019350.1 XP_016874839.1 5 847
          XM_017019351.1 XP_016874840.1 5 1013
          XM_017019352.1 XP_016874841.1 6 1034
          XM_017019353.1 XP_016874842.1 4 726
          XM_017019354.1 XP_016874843.1 4 892
          GenBank mRNA AF044215.1 - 3 221
          AF113226.1 - 2 192
          AF161480.1 - 3 205
          AF180473.1 - 3 200
          AK000662.1 - 3 230
          AK095789.1 - 3 752
          AK307479.1 - 4 1013
          AK315715.1 - 4 537
          BC002597.2 - 3 211 3143289
          BC011826.2 - 3 219 3832035

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001011988, XM_006241366, XM_006241365, XM_006241367, XM_006241363, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001037848, NM_001037846, NM_001037847, AK078121, BC065171, and others.

          Panther Classification:

          Molecular Function -

          general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
          nuclear-transcribed mRNA poly(A) tail shortening
          trophectodermal cell differentiation
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          positive regulation of cytoplasmic mRNA processing body assembly
          negative regulation of translation
          gene silencing by RNA
          negative regulation of intracellular estrogen receptor signaling pathway
          RNA phosphodiester bond hydrolysis, exonucleolytic
          regulation of stem cell population maintenance
          RNA polymerase II transcription cofactor activity
          RNA polymerase II transcription corepressor binding
          poly(A)-specific ribonuclease activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0