Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CYLD SIRNA Assays ›
          Gene Symbol:
          CYLD
          Gene Name:
          CYLD lysine 63 deubiquitinase
          Gene Aliases:
          BRSS, CDMT, CYLD1, CYLDI, EAC, MFT, MFT1, SBS, TEM, USPL2
          Chromosome Location:
          Chr.16: 50742026 - 50801935 on Build GRCh38
          UniGene ID:
          Hs.578973
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 43399

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CYLD
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042355.1 NP_001035814.1 3 397
          NM_001042412.1 NP_001035877.1 3 421
          NM_015247.2 NP_056062.1 4 534
          XM_005255812.2 XP_005255869.1 4 494
          XM_006721149.1 XP_006721212.1 4 558
          XM_011522907.2 XP_011521209.1 3 320
          XM_011522908.1 XP_011521210.1 3 412
          XM_011522911.2 XP_011521213.1 3 412
          XM_017022977.1 XP_016878466.1 3 692
          XM_017022978.1 XP_016878467.1 3 488
          XM_017022979.1 XP_016878468.1 3 448
          XM_017022980.1 XP_016878469.1 3 481
          XM_017022981.1 XP_016878470.1 3 379
          GenBank mRNA AF161542.1 - 2 368
          AJ250014.1 - 4 510
          AK056226.1 - 3 352
          AK225202.1 - 4 514
          AK292975.1 - 3 379
          AK310832.1 - 3 369
          BC012342.1 - 3 395 4552767
          BI458962.1 - 3 392

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquitin-dependent protein catabolic process
          cell cycle
          regulation of mitotic cell cycle
          regulation of tumor necrosis factor-mediated signaling pathway
          Wnt signaling pathway
          negative regulation of NF-kappaB transcription factor activity
          negative regulation of type I interferon production
          negative regulation of NF-kappaB import into nucleus
          negative regulation of T cell differentiation
          necroptotic process
          nucleotide-binding oligomerization domain containing signaling pathway
          regulation of microtubule cytoskeleton organization
          protein K63-linked deubiquitination
          negative regulation of canonical Wnt signaling pathway
          ripoptosome assembly involved in necroptotic process
          regulation of cilium assembly
          positive regulation of extrinsic apoptotic signaling pathway
          regulation of intrinsic apoptotic signaling pathway
          thiol-dependent ubiquitin-specific protease activity
          protein binding
          zinc ion binding
          protein kinase binding
          Lys63-specific deubiquitinase activity
          proline-rich region binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0