Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTPRK SIRNA Assays ›
          Gene Symbol:
          PTPRK
          Gene Name:
          protein tyrosine phosphatase, receptor type K
          Gene Aliases:
          R-PTP-kappa
          Chromosome Location:
          Chr.6: 127968779 - 128520847 on Build GRCh38
          UniGene ID:
          Hs.155919
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 4370

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTPRK
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135648.2 NP_001129120.1 2 471
          NM_001291981.1 NP_001278910.1 2 471
          NM_001291982.1 NP_001278911.1 2 471
          NM_001291983.1 NP_001278912.1 2 471
          NM_001291984.1 NP_001278913.1 2 471
          NM_002844.3 NP_002835.2 2 471
          XM_006715537.2 XP_006715600.1 2 522
          XM_011536014.2 XP_011534316.1 2 522
          XM_011536015.2 XP_011534317.1 2 522
          XM_011536016.2 XP_011534318.1 2 164
          XM_011536017.2 XP_011534319.1 3 762
          XM_011536018.2 XP_011534320.1 3 435
          XM_011536020.2 XP_011534322.1 2 522
          XM_011536021.2 XP_011534323.1 2 522
          XM_017011145.1 XP_016866634.1 3 486
          XM_017011146.1 XP_016866635.1 3 435
          XM_017011147.1 XP_016866636.1 3 416
          XM_017011148.1 XP_016866637.1 3 413
          GenBank mRNA AF533875.1 - 2 236
          AK295028.1 - 2 396
          BC063596.1 - 3 480
          BC140775.1 - 2 273 9021643
          BC144512.1 - 2 273 9053039
          BC144513.1 - 2 273 9053040
          BI755683.1 - 375
          CR749277.1 - 2 412
          DB020920.1 - 2 472
          L77886.1 - 2 170
          Z70660.1 - 2 155

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          extracellular matrix glycoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein dephosphorylation
          cell adhesion
          signal transduction
          transforming growth factor beta receptor signaling pathway
          negative regulation of cell proliferation
          negative regulation of keratinocyte proliferation
          cell migration
          negative regulation of cell migration
          neuron projection development
          protein localization to cell surface
          cellular response to reactive oxygen species
          cellular response to UV
          peptidyl-tyrosine dephosphorylation
          negative regulation of cell cycle
          negative regulation of transcription, DNA-templated
          focal adhesion assembly
          protein tyrosine phosphatase activity
          transmembrane receptor protein tyrosine phosphatase activity
          protein binding
          beta-catenin binding
          protein kinase binding
          gamma-catenin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0