Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SOX5 SIRNA Assays ›
          Gene Symbol:
          SOX5
          Gene Name:
          SRY-box 5
          Gene Aliases:
          L-SOX5, L-SOX5B, L-SOX5F, LAMSHF
          Chromosome Location:
          Chr.12: 23529499 - 24562701 on Build GRCh38
          UniGene ID:
          Hs.657542
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 44023

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SOX5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001261415.1 NP_001248344.1 11, 12 1617
          NM_006940.4 NP_008871.3 11, 12 1593
          NM_152989.3 NP_694534.1 14, 15 1826
          NM_178010.2 NP_821078.1 3, 4 393
          XM_011520831.2 XP_011519133.1 11, 12 1455
          XM_011520832.2 XP_011519134.1 11, 12 1567
          XM_011520833.2 XP_011519135.1 11, 12 1896
          XM_011520834.2 XP_011519136.1 15, 16 2029
          XM_011520835.2 XP_011519137.1 14, 15 2081
          XM_011520837.2 XP_011519139.1 11, 12 1638
          XM_011520838.2 XP_011519140.1 11, 12 1458
          XM_011520842.2 XP_011519144.1 4, 5 518
          XM_017019888.1 XP_016875377.1 12, 13 1910
          XM_017019889.1 XP_016875378.1 12, 13 1906
          XM_017019890.1 XP_016875379.1 15, 16 2045
          XM_017019891.1 XP_016875380.1 15, 16 1903
          XM_017019892.1 XP_016875381.1 16, 17 2124
          XM_017019893.1 XP_016875382.1 13, 14 1984
          XM_017019894.1 XP_016875383.1 11, 12 3914
          XM_017019895.1 XP_016875384.1 11, 12 1481
          XM_017019896.1 XP_016875385.1 15, 16 2042
          XM_017019897.1 XP_016875386.1 15, 16 1940
          XM_017019898.1 XP_016875387.1 15, 16 1937
          XM_017019899.1 XP_016875388.1 14, 15 1649
          XM_017019900.1 XP_016875389.1 11, 12 2263
          XM_017019901.1 XP_016875390.1 11, 12 6719
          GenBank mRNA AB081588.1 - 14, 15 1824
          AB081589.1 - 11, 12 1593
          AK074317.1 - 11, 12 1520
          AK096569.1 - 11, 12 1887
          AK098610.1 - 3, 4 393
          AK223536.1 - 3, 4 393
          AK303970.1 - 11, 12 1617
          BC029220.2 - 2, 3 331
          BC035227.2 - 2, 3 331
          BC047665.2 - 2, 3 331
          CR456713.1 - 2, 3 243
          JX570584.1 - 12, 13 1638
          JX570585.1 - 12, 13 1533
          S83308.1 - 2, 3 327

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001271267, XM_008763350, XM_006237604, XM_008763349, BC086415.
          This siRNA also targets the following transcripts in Mouse: NM_001243163, NM_001113559, NM_011444, X65657, AK015212, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          positive regulation of chondrocyte differentiation
          asymmetric neuroblast division
          positive regulation of cartilage development
          cellular response to transforming growth factor beta stimulus
          positive regulation of mesenchymal stem cell differentiation
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0