Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CBLB SIRNA Assays ›
          Gene Symbol:
          CBLB
          Gene Name:
          Cbl proto-oncogene B
          Gene Aliases:
          Cbl-b, Nbla00127, RNF56
          Chromosome Location:
          Chr.3: 105655461 - 105869552 on Build GRCh38
          UniGene ID:
          Hs.430589
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 44069

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CBLB
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321786.1 NP_001308715.1 18 2864
          NM_001321788.1 NP_001308717.1 18 2783
          NM_001321789.1 NP_001308718.1 17 2655
          NM_001321790.1 NP_001308719.1 17 2605
          NM_001321791.1 NP_001308720.1 17 2651
          NM_001321793.1 NP_001308722.1 17 2827
          NM_001321794.1 NP_001308723.1 17 2636
          NM_001321795.1 NP_001308724.1 17 2812
          NM_001321796.1 NP_001308725.1 17 2636
          NM_001321797.1 NP_001308726.1 16 2504
          NM_001321798.1 NP_001308727.1 16 2680
          NM_001321799.1 NP_001308728.1 16 2680
          NM_001321806.1 NP_001308735.1 17 2626
          NM_001321807.1 NP_001308736.1 19 3103
          NM_001321808.1 NP_001308737.1 18 2795
          NM_001321811.1 NP_001308740.1 16 2655
          NM_001321813.1 NP_001308742.1 18 2693
          NM_001321816.1 NP_001308745.1 15 2523
          NM_001321820.1 NP_001308749.1 19 2959
          NM_001321822.1 NP_001308751.1 19 3009
          NM_170662.4 NP_733762.2 18 2959
          NR_135806.1 20 3184
          NR_135807.1 20 3030
          NR_135808.1 19 2985
          NR_135809.1 18 2941
          NR_135810.1 20 2886
          NR_135811.1 19 2806
          NR_135812.1 18 2665
          XM_011513257.1 XP_011511559.1 18 2779
          XM_017007395.1 XP_016862884.1 18 2731
          XM_017007396.1 XP_016862885.1 17 2770
          XM_017007397.1 XP_016862886.1 17 2756
          XM_017007398.1 XP_016862887.1 17 2582
          XM_017007399.1 XP_016862888.1 17 2807
          XM_017007400.1 XP_016862889.1 16 2624
          GenBank mRNA AB075490.1 - 7 1242
          AK094184.1 - 9 1185
          AK123147.1 - 3 616
          AK130668.1 - 1 1176
          AK292792.1 - 18 2900
          AK302533.1 - 17 2605
          AK316276.1 - 17 2585
          BC032851.2 - 18 2900 5269036
          BX649066.1 - 8 875
          CR749655.1 - 6 654
          DQ349203.1 - 17 2637
          DQ349204.1 - 17 2637
          U26710.1 - 18 2959

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          damaged DNA-binding protein ligase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          NLS-bearing protein import into nucleus
          signal transduction
          cell surface receptor signaling pathway
          response to gravity
          response to hormone
          regulation of signaling
          phosphotyrosine binding
          ubiquitin-protein transferase activity
          signal transducer activity
          calcium ion binding
          protein binding
          zinc ion binding
          ligase activity
          protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0