Search
Search

FAM171B| Gene Symbol: FAM171B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177454.3 | NP_803237.3 | 7 | 1138 | ||
| GenBank mRNA | AB075826.1 | - | 6 | 587 | ||
| AK091423.1 | - | 7 | 1141 | |||
| AK095604.1 | - | 4 | 420 | |||
| AK289737.1 | - | 7 | 1141 | |||
| BC060872.1 | - | 7 | 1144 | 30347998 | ||