Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SMPD4 SIRNA Assays ›
          Gene Symbol:
          SMPD4 LOC150776
          Gene Name:
          sphingomyelin phosphodiesterase 4
          sphingomyelin phosphodiesterase 4, neutral membrane (neutral sphingomyelinase-3) pseudogene
          Gene Aliases:
          NET13, NSMASE-3, NSMASE3
          -
          Chromosome Location:
          Chr.2: 130151392 - 130182750 on Build GRCh38
          Chromosome Location:
          Chr.2: 131492813 - 131521576 on Build GRCh38
          UniGene ID:
          Hs.516450
          Hs.535622
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 44448

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SMPD4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001171083.2 NP_001164554.1 8 893
          NM_017751.4 NP_060221.2 10 1112
          NM_017951.4 NP_060421.2 10 1112
          NR_033230.2 10 2105
          NR_033231.2 9 969
          NR_033232.2 9 1036
          XM_011511443.2 XP_011509745.1 10 1274
          XM_011511444.2 XP_011509746.1 10 1273
          XM_011511445.2 XP_011509747.1 10 968
          XM_017004448.1 XP_016859937.1 10 1014
          XM_017004449.1 XP_016859938.1 10 1014
          GenBank mRNA AB037839.1 - 10 2105
          AK000304.1 - 10 949
          AK000763.1 - 5 442
          AK056501.1 - 10 904
          AK297257.1 - 4 298
          AK298609.1 - 4 336
          AK299185.1 - 8 743
          AK301576.1 - 4 331
          AK303427.1 - 4 330
          AK303519.1 - 9 873
          BC040941.1 - 3 317
          BC064947.1 - 9 791 6023304
          EU367941.1 - 10 956
          Gene Symbol: LOC150776
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_026922.1 10 1209
          GenBank mRNA AK293811.1 - 10 946
          AK300337.1 - 8 731
          AK303293.1 - 8 779
          AK304811.1 - 9 1007
          AK308371.1 - 2 114
          BC093080.1 - 10 932
          CN337608.1 - 1 46

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          hydrolase phosphodiesterase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          sphingomyelin catabolic process
          glycosphingolipid metabolic process
          glycerophospholipid catabolic process
          ceramide biosynthetic process
          cellular response to tumor necrosis factor
          sphingomyelin phosphodiesterase activity
          metal ion binding
          sphingomyelin phosphodiesterase D activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline