Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IFFO1 SIRNA Assays ›
          Gene Symbol:
          IFFO1
          Gene Name:
          intermediate filament family orphan 1
          Gene Aliases:
          HOM-TES-103, IFFO
          Chromosome Location:
          Chr.12: 6539528 - 6556102 on Build GRCh38
          UniGene ID:
          Hs.15243
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 44568

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IFFO1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039670.2 NP_001034759.1 6, 7 1303
          NM_001193457.1 NP_001180386.1 6, 7 1327
          NM_080730.4 NP_542768.2 6, 7 1303
          NR_036467.1 6, 7 1400
          XM_006718975.3 XP_006719038.1 6, 7 1319
          XM_006718976.3 XP_006719039.1 5, 6 1313
          XM_006718977.3 XP_006719040.1 5, 6 1313
          XM_006718981.3 XP_006719044.1 7, 8 988
          XM_011520947.2 XP_011519249.1 6, 7 1322
          XM_011520948.2 XP_011519250.1 6, 7 1319
          XM_011520949.2 XP_011519251.1 5, 6 1313
          XM_011520952.2 XP_011519254.1 7, 8 988
          XM_017019169.1 XP_016874658.1 6, 7 1322
          XM_017019170.1 XP_016874659.1 6, 7 1125
          XM_017019171.1 XP_016874660.1 6, 7 1319
          XM_017019172.1 XP_016874661.1 7, 8 991
          XM_017019173.1 XP_016874662.1 6, 7 622
          GenBank mRNA AF124432.1 - 6, 7 964
          AK000710.1 - 1, 2 536
          AK095106.1 - 2, 3 881
          AK298905.1 - 6, 7 536
          BC001790.1 - 6, 7 1380
          BC002857.2 - 5, 6 441
          BC004384.1 - 5, 6 441
          BC010431.1 - 6, 7 1380
          BC023664.2 - 6, 7 1380
          BC063004.1 - 6, 7 578
          BC071170.1 - 1, 2 452
          BC103817.1 - 1, 2 457
          BC103818.1 - 1, 2 457
          BC110387.1 - 6, 7 1293 5183989
          BC113848.1 - 1, 2 457
          BC114454.1 - 1, 2 457
          BM922290.1 - 476
          BX327856.2 - 6, 7 519
          BX648701.1 - 7, 8 889
          BX648743.1 - 7, 8 889
          DA268043.1 - 1, 2 518

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001108647, XM_006237373, XM_006237372, XM_008763292, XM_006237376, and others.
          This siRNA also targets the following transcripts in Mouse: AK044709, AK028273, AK189508, BC057601, AK053568, and others.

          Panther Classification:

          Molecular Function -

          dehydrogenase oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Process(es)

          structural molecule activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0