Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATXN2L SIRNA Assays ›
Gene Symbol:
ATXN2L
Gene Name:
ataxin 2 like
Gene Aliases:
A2D, A2LG, A2LP, A2RP
Chromosome Location:
Chr.16: 28823048 - 28837237 on Build GRCh38
UniGene ID:
Hs.460499
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
44585
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATXN2L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308230.1
NP_001295159.1
3
585
NM_007245.3
NP_009176.2
3
585
NM_017492.3
NP_059867.3
3
585
NM_145714.2
NP_663760.1
3
585
NM_148414.2
NP_680780.1
3
585
NM_148415.2
NP_680781.1
3
585
NM_148416.2
NP_680782.1
3
585
XM_005255061.1
XP_005255118.1
3
585
XM_005255062.1
XP_005255119.1
3
585
XM_005255063.3
XP_005255120.1
3
585
XM_005255064.1
XP_005255121.1
3
585
XM_005255065.1
XP_005255122.1
3
585
XM_005255066.1
XP_005255123.1
3
585
XM_005255067.1
XP_005255124.1
3
585
XM_005255068.1
XP_005255125.1
3
585
XM_005255069.1
XP_005255126.1
3
585
XM_005255070.1
XP_005255127.1
3
585
XM_005255071.1
XP_005255128.1
3
585
XM_005255074.1
XP_005255131.1
3
585
XM_005255075.1
XP_005255132.1
3
585
XM_005255076.1
XP_005255133.1
3
585
XM_005255077.1
XP_005255134.1
3
468
XM_006721007.1
XP_006721070.1
3
585
XM_006721008.1
XP_006721071.1
3
585
XM_006721009.1
XP_006721072.1
3
585
XM_006721010.2
XP_006721073.1
3
585
XM_006721011.1
XP_006721074.1
3
585
XM_006721012.3
XP_006721075.1
3
585
XM_011545719.2
XP_011544021.1
3
585
XM_011545721.2
XP_011544023.1
3
585
XM_011545722.1
XP_011544024.1
3
285
XM_017022891.1
XP_016878380.1
3
468
XM_017022892.1
XP_016878381.1
3
285
GenBank mRNA
AF034373.1
-
5
687
AJ317970.1
-
3
541
AJ317971.2
-
3
541
AJ317972.2
-
3
541
AJ317973.2
-
3
541
AJ317974.2
-
3
541
AK289981.1
-
3
585
AY188334.1
-
5
687
AY188335.1
-
5
687
AY188336.1
-
5
687
AY188337.1
-
5
687
AY188338.1
-
5
687
BC068012.1
-
4
443
BC082760.1
-
3
381
5812230
BC136584.1
-
3
538
9020574
U70671.1
-
3
215
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cytoplasmic mRNA processing body assembly
stress granule assembly
cell-cell adhesion
protein binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0