Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IL17RC SIRNA Assays ›
Gene Symbol:
IL17RC
Gene Name:
interleukin 17 receptor C
Gene Aliases:
CANDF9, IL17-RL, IL17RL
Chromosome Location:
Chr.3: 9917074 - 9933627 on Build GRCh38
UniGene ID:
Hs.129959
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
44651
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IL17RC
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001203263.1
NP_001190192.1
6
789
NM_001203264.1
NP_001190193.1
6
789
NM_001203265.1
NP_001190194.1
6
789
NM_032732.5
NP_116121.2
6
789
NM_153460.3
NP_703190.1
6
789
NM_153461.3
NP_703191.1
6
978
NR_037807.1
6
789
XM_017007326.1
XP_016862815.1
6
978
XM_017007327.1
XP_016862816.1
6
978
XM_017007328.1
XP_016862817.1
6
978
XM_017007329.1
XP_016862818.1
6
978
XM_017007330.1
XP_016862819.1
6
978
XM_017007331.1
XP_016862820.1
6
978
XM_017007332.1
XP_016862821.1
6
978
XM_017007333.1
XP_016862822.1
6
978
XM_017007334.1
XP_016862823.1
5
903
XM_017007335.1
XP_016862824.1
6
978
XM_017007336.1
XP_016862825.1
6
978
XM_017007337.1
XP_016862826.1
5
903
XM_017007338.1
XP_016862827.1
6
978
XM_017007339.1
XP_016862828.1
6
978
XM_017007340.1
XP_016862829.1
6
766
XM_017007341.1
XP_016862830.1
6
766
XM_017007342.1
XP_016862831.1
5
690
XM_017007343.1
XP_016862832.1
5
690
XM_017007344.1
XP_016862833.1
6
766
XM_017007345.1
XP_016862834.1
6
766
XM_017007346.1
XP_016862835.1
6
766
XM_017007347.1
XP_016862836.1
5
690
XM_017007348.1
XP_016862837.1
6
766
XM_017007349.1
XP_016862838.1
5
690
GenBank mRNA
AF458065.1
-
6
744
AK075534.1
-
6
754
AK092907.1
-
6
979
AK096824.1
-
6
814
AK225650.1
-
6
734
AK290670.1
-
6
826
AK294616.1
-
5
632
AK295377.1
-
5
691
AK303171.1
-
5
600
AK308241.1
-
5
674
AK315024.1
-
6
766
AK315053.1
-
6
734
AY358840.1
-
6
751
AY359098.1
-
6
779
EF676032.1
-
6
700
EF676033.1
-
6
547
EF676034.1
-
6
558
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
cytokine-mediated signaling pathway
defense response to fungus
granulocyte chemotaxis
positive regulation of cytokine production involved in inflammatory response
positive regulation of interleukin-6 secretion
receptor binding
interleukin-17 receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0