Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SEMA6D SIRNA Assays ›
          Gene Symbol:
          SEMA6D
          Gene Name:
          semaphorin 6D
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 47184101 - 47774223 on Build GRCh38
          UniGene ID:
          Hs.511265
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 44655

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SEMA6D
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001198999.1 NP_001185928.1 7, 8 727
          NM_020858.1 NP_065909.1 4, 5 715
          NM_024966.2 NP_079242.2 4, 5 715
          NM_153616.1 NP_705869.1 4, 5 715
          NM_153617.1 NP_705870.1 4, 5 715
          NM_153618.1 NP_705871.1 4, 5 715
          NM_153619.1 NP_705872.1 4, 5 715
          XM_005254685.3 XP_005254742.1 4, 5 677
          XM_005254686.2 XP_005254743.1 4, 5 621
          XM_005254687.2 XP_005254744.1 4, 5 621
          XM_005254689.3 XP_005254746.1 4, 5 677
          XM_011522075.2 XP_011520377.1 7, 8 832
          XM_011522076.2 XP_011520378.1 4, 5 621
          XM_011522077.2 XP_011520379.1 4, 5 621
          XM_011522078.2 XP_011520380.1 4, 5 621
          XM_011522079.2 XP_011520381.1 4, 5 621
          XM_011522080.2 XP_011520382.1 4, 5 621
          XM_011522081.2 XP_011520383.1 4, 5 621
          XM_017022617.1 XP_016878106.1 7, 8 645
          XM_017022618.1 XP_016878107.1 4, 5 677
          XM_017022619.1 XP_016878108.1 4, 5 677
          XM_017022620.1 XP_016878109.1 7, 8 832
          XM_017022621.1 XP_016878110.1 7, 8 832
          GenBank mRNA AB040912.2 - 7, 8 727
          AF389426.1 - 4, 5 715
          AF389427.1 - 4, 5 715
          AF389428.1 - 4, 5 715
          AF389429.1 - 4, 5 715
          AF389430.1 - 4, 5 715
          AK021660.1 - 1, 2 13
          BC150253.1 - 7, 8 727 9007179

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane-bound signaling molecule intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          neural crest cell migration
          positive regulation of smooth muscle cell migration
          negative regulation of smooth muscle cell migration
          ventricular system development
          positive regulation of cell migration
          negative regulation of axon extension involved in axon guidance
          negative chemotaxis
          semaphorin-plexin signaling pathway
          semaphorin receptor binding
          chemorepellent activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0