Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRKRA SIRNA Assays ›
          Gene Symbol:
          PRKRA
          Gene Name:
          protein activator of interferon induced protein kinase EIF2AK2
          Gene Aliases:
          DYT16, HSD14, PACT, RAX
          Chromosome Location:
          Chr.2: 178431414 - 178451240 on Build GRCh38
          UniGene ID:
          Hs.570274
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 449
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price 6,301.00
          Your Price
          Online offer:
          6301.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PRKRA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001139517.1 NP_001132989.1 1 383
          NM_001139518.1 NP_001132990.1 2 104
          NM_001316362.1 NP_001303291.1 2 312
          NM_003690.4 NP_003681.1 2 303
          XM_011512063.1 XP_011510365.1 2 185
          XM_011512066.1 XP_011510368.1 2 349
          XM_017005159.1 XP_016860648.1 2 185
          GenBank mRNA AF072860.1 - 2 198
          AF083033.1 - 2 194
          AK223107.1 - 2 202
          AK290601.1 - 2 229
          AK296019.1 - 2 223
          AL136615.1 - 2 227
          AL833867.1 - 1 383
          AY251164.1 - 2 121
          BC009470.1 - 2 209 3503747
          BG827125.1 - 2 304
          BT007243.1 - 2 102
          CR533525.1 - 2 102
          HY049565.1 - 2 312

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00269379_m1

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          immune response
          negative regulation of cell proliferation
          response to virus
          production of siRNA involved in RNA interference
          gene silencing by RNA
          pre-miRNA processing
          cellular response to oxidative stress
          production of miRNAs involved in gene silencing by miRNA
          outer ear morphogenesis
          middle ear morphogenesis
          positive regulation of catalytic activity
          skeletal system morphogenesis
          positive regulation of intrinsic apoptotic signaling pathway
          double-stranded RNA binding
          protein binding
          enzyme activator activity
          enzyme binding
          identical protein binding
          protein homodimerization activity
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0