Search
Search

HAS3| Gene Symbol: HAS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199280.1 | NP_001186209.1 | 2 | 774 | ||
| NM_005329.2 | NP_005320.2 | 2 | 706 | |||
| NM_138612.2 | NP_619515.1 | 2 | 641 | |||
| XM_005255921.2 | XP_005255978.1 | 2 | 652 | |||
| XM_011523061.2 | XP_011521363.1 | 3 | 774 | |||
| GenBank mRNA | AF232772.1 | - | 1 | 550 | ||
| AF234839.1 | - | 2 | 706 | |||
| AK291400.1 | - | 2 | 774 | |||
| BC021853.1 | - | 2 | 610 | 4477285 | ||