Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CDC14B SIRNA Assays ›
Gene Symbol:
CDC14B
Gene Name:
cell division cycle 14B
Gene Aliases:
CDC14B3, Cdc14B1, Cdc14B2, hCDC14B
Chromosome Location:
Chr.9: 96492738 - 96619830 on Build GRCh38
UniGene ID:
Hs.40582
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
45559
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CDC14B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077181.1
NP_001070649.1
2, 3
251
NM_003671.3
NP_003662.1
2, 3
709
NM_033331.2
NP_201588.1
2, 3
709
XM_005252288.3
XP_005252345.1
2, 3
306
XM_011519147.2
XP_011517449.1
2, 3
656
XM_011519148.2
XP_011517450.1
2, 3
650
XM_011519149.2
XP_011517451.1
2, 3
654
XM_011519150.2
XP_011517452.1
2, 3
653
XM_011519151.2
XP_011517453.1
2, 3
653
XM_011519152.2
XP_011517454.1
2, 3
656
XM_011519153.2
XP_011517455.1
2, 3
652
XM_011519156.2
XP_011517458.2
2, 3
827
XM_011519158.1
XP_011517460.1
2, 3
325
XM_011519159.2
XP_011517461.2
2, 3
123
XM_017015240.1
XP_016870729.1
2, 3
656
XM_017015241.1
XP_016870730.1
2, 3
661
XM_017015242.1
XP_016870731.1
2, 3
325
XM_017015243.1
XP_016870732.1
2, 3
325
XM_017015244.1
XP_016870733.1
2, 3
306
XM_017015245.1
XP_016870734.1
2, 3
827
XM_017015246.1
XP_016870735.1
2, 3
306
XM_017015247.1
XP_016870736.1
2, 3
306
XM_017015248.1
XP_016870737.1
2, 3
272
XM_017015249.1
XP_016870738.1
2, 3
215
GenBank mRNA
AF023158.1
-
2, 3
257
AF064104.1
-
2, 3
709
AF064105.2
-
2, 3
709
AK126388.1
-
2, 3
243
AK225361.1
-
2, 3
242
DA943563.1
-
2, 3
251
EF611343.1
-
2, 3
257
Back To Top
More Information
Panther Classification:
Molecular Function -
protein phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
DNA repair
protein dephosphorylation
G2 DNA damage checkpoint
peptidyl-tyrosine dephosphorylation
positive regulation of ubiquitin protein ligase activity
protein serine/threonine phosphatase activity
protein tyrosine phosphatase activity
protein binding
protein tyrosine/serine/threonine phosphatase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0