Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TBX1 SIRNA Assays ›
          Gene Symbol:
          TBX1
          Gene Name:
          T-box 1
          Gene Aliases:
          CAFS, CATCH22, CTHM, DGCR, DGS, DORV, TBX1C, TGA, VCF, VCFS
          Chromosome Location:
          Chr.22: 19756703 - 19783593 on Build GRCh38
          UniGene ID:
          Hs.173984
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 45666

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TBX1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005992.1 NP_005983.1 4 570
          NM_080646.1 NP_542377.1 4 570
          NM_080647.1 NP_542378.1 4 570
          XM_006724312.2 XP_006724375.1 4 739
          XM_017028925.1 XP_016884414.1 3 615
          XM_017028926.1 XP_016884415.1 4 705
          XM_017028928.1 XP_016884417.1 3 614
          GenBank mRNA AF012130.1 - 4 570
          AF012131.1 - 4 570
          AF373867.1 - 4 570

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          Rel homology transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          blood vessel development
          cell fate specification
          neural crest cell migration
          positive regulation of protein phosphorylation
          lymph vessel development
          positive regulation of mesenchymal cell proliferation
          heart morphogenesis
          outflow tract septum morphogenesis
          outflow tract morphogenesis
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          determination of left/right symmetry
          pattern specification process
          mesoderm development
          heart development
          muscle organ development
          sensory perception of sound
          cell proliferation
          positive regulation of cell proliferation
          anterior/posterior pattern specification
          vagus nerve morphogenesis
          epithelial cell differentiation
          thyroid gland development
          social behavior
          aorta morphogenesis
          ear morphogenesis
          inner ear morphogenesis
          outer ear morphogenesis
          middle ear morphogenesis
          odontogenesis of dentin-containing tooth
          muscle cell fate commitment
          positive regulation of MAPK cascade
          tongue morphogenesis
          cellular response to fibroblast growth factor stimulus
          negative regulation of cell differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          retinoic acid receptor signaling pathway
          blood vessel morphogenesis
          thymus development
          muscle organ morphogenesis
          embryonic cranial skeleton morphogenesis
          embryonic viscerocranium morphogenesis
          semicircular canal morphogenesis
          artery morphogenesis
          positive regulation of epithelial cell proliferation
          parathyroid gland development
          soft palate development
          pharyngeal system development
          face morphogenesis
          muscle tissue morphogenesis
          coronary artery morphogenesis
          enamel mineralization
          cellular response to retinoic acid
          cochlea morphogenesis
          mesenchymal cell apoptotic process
          regulation of organ morphogenesis
          positive regulation of tongue muscle cell differentiation
          negative regulation of mesenchymal cell apoptotic process
          transcription factor activity, sequence-specific DNA binding
          protein homodimerization activity
          sequence-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zcvn6:80/100.66.79.31:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0