Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRPF4B SIRNA Assays ›
Gene Symbol:
PRPF4B
Gene Name:
pre-mRNA processing factor 4B
Gene Aliases:
PR4H, PRP4, PRP4H, PRP4K, dJ1013A10.1
Chromosome Location:
Chr.6: 4019402 - 4064983 on Build GRCh38
UniGene ID:
Hs.159014
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
461
Purification
In-Vivo Ready
Standard
HPLC
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRPF4B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003913.4
NP_003904.3
5, 6
1733
XM_011514970.2
XP_011513272.1
5, 6
3666
XM_017011410.1
XP_016866899.1
5, 6
3666
XM_017011411.1
XP_016866900.1
5, 6
3666
XM_017011412.1
XP_016866901.1
5, 6
3666
XM_017011413.1
XP_016866902.1
5, 6
3666
XM_017011414.1
XP_016866903.1
5, 6
3666
XM_017011415.1
XP_016866904.1
5, 6
3666
XM_017011416.1
XP_016866905.1
5, 6
3666
XM_017011417.1
XP_016866906.1
5, 6
3666
XM_017011418.1
XP_016866907.1
5, 6
3666
XM_017011419.1
XP_016866908.1
5, 6
3666
XM_017011420.1
XP_016866909.1
5, 6
3666
XM_017011421.1
XP_016866910.1
5, 6
3666
XM_017011422.1
XP_016866911.1
5, 6
3666
XM_017011423.1
XP_016866912.1
5, 6
3666
XM_017011424.1
XP_016866913.1
5, 6
3666
XM_017011425.1
XP_016866914.1
5, 6
3666
XM_017011426.1
XP_016866915.1
5, 6
3666
XM_017011427.1
XP_016866916.1
5, 6
3666
XM_017011428.1
XP_016866917.1
5, 6
3666
XM_017011429.1
XP_016866918.1
5, 6
3666
XM_017011430.1
XP_016866919.1
5, 6
3666
XM_017011431.1
XP_016866920.1
5, 6
3666
XM_017011432.1
XP_016866921.1
5, 6
3666
XM_017011433.1
XP_016866922.1
5, 6
3666
XM_017011434.1
XP_016866923.1
5, 6
3666
XM_017011435.1
XP_016866924.1
5, 6
3666
GenBank mRNA
AB011108.1
-
5, 6
1706
AF283465.1
-
5, 6
1684
AK291244.1
-
5, 6
1711
AK303047.1
-
5, 6
1758
AY029347.1
-
5, 6
1642
BC009844.2
-
3, 4
381
BC034969.1
-
5, 6
1746
4821304
U48736.1
-
2, 3
222
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
protein phosphorylation
RNA splicing
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0