Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ECHDC2 SIRNA Assays ›
Gene Symbol:
ECHDC2
Gene Name:
enoyl-CoA hydratase domain containing 2
Gene Aliases:
-
Chromosome Location:
Chr.1: 52895910 - 52927212 on Build GRCh38
UniGene ID:
Hs.476319
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
46287
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ECHDC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198961.1
NP_001185890.1
2
241
NM_001198962.1
NP_001185891.1
2
241
NM_001319958.1
NP_001306887.1
3
754
NM_018281.3
NP_060751.2
2
241
XM_011541709.2
XP_011540011.1
3
452
XM_011541710.2
XP_011540012.1
3
452
XM_011541711.2
XP_011540013.1
3
452
XM_011541712.2
XP_011540014.1
3
452
XM_011541713.2
XP_011540015.1
2
4229
XM_011541715.2
XP_011540017.1
3
677
XM_011541719.2
XP_011540021.1
3
1674
XM_011541720.2
XP_011540022.1
2
3998
XM_011541721.2
XP_011540023.1
4
691
XM_011541722.2
XP_011540024.1
3
309
XM_011541723.2
XP_011540025.1
3
409
XM_011541724.2
XP_011540026.1
3
452
XM_011541725.2
XP_011540027.1
3
452
XM_011541726.2
XP_011540028.1
2
4229
XM_011541727.2
XP_011540029.1
2
3998
XM_017001638.1
XP_016857127.1
4
2063
XM_017001639.1
XP_016857128.1
3
468
XM_017001640.1
XP_016857129.1
2
631
XM_017001641.1
XP_016857130.1
4
1832
XM_017001642.1
XP_016857131.1
2
4229
GenBank mRNA
AK001810.1
-
2
193
AK299834.1
-
3
410
AK300896.1
-
3
280
AK302285.1
-
5
820
AK307381.1
-
2
136
AK310650.1
-
3
373
AK315906.1
-
3
372
BC038389.1
-
3
672
BC044574.1
-
2
171
5314083
BC051887.1
-
2
229
CF127686.1
-
2
185
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001106675.
This siRNA also targets the following transcripts in Mouse: NM_026728, AK166965, AK153878.
Panther Classification:
Molecular Function -
hydratase
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid beta-oxidation
enoyl-CoA hydratase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0