Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCL3 SIRNA Assays ›
Gene Symbol:
Ccl3
Gene Name:
chemokine (C-C motif) ligand 3
Gene Aliases:
MIP-1a, Scya3
Chromosome Location:
Chr.10: 70869516 - 70871066 on Build Rnor_6.0
UniGene ID:
Rn.10139
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
46699
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ccl3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_013025.2
NP_037157.2
3
309
GenBank mRNA
EF121994.1
-
3
240
EF121995.1
-
3
240
U06435.1
-
3
240
U22414.1
-
3
309
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_011337, AF065942, AF065939, BX524012, AK150590, and others.
Panther Classification:
Molecular Function -
cytokine
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
osteoblast differentiation
cell activation
monocyte chemotaxis
protein phosphorylation
calcium ion transport
cellular calcium ion homeostasis
exocytosis
chemotaxis
inflammatory response
cytoskeleton organization
positive regulation of cytosolic calcium ion concentration
cell-cell signaling
regulation of cell shape
response to toxic substance
positive regulation of gene expression
negative regulation of gene expression
T cell chemotaxis
release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
calcium-mediated signaling
signaling
positive regulation of cell migration
negative regulation of bone mineralization
neutrophil chemotaxis
leukocyte chemotaxis
lipopolysaccharide-mediated signaling pathway
positive regulation of tumor necrosis factor production
response to drug
positive regulation of catalytic activity
eosinophil degranulation
protein kinase B signaling
positive regulation of neuron apoptotic process
astrocyte cell migration
negative regulation by host of viral transcription
negative regulation of osteoclast differentiation
positive regulation of osteoclast differentiation
positive regulation of transcription from RNA polymerase II promoter
eosinophil chemotaxis
macrophage chemotaxis
lymphocyte chemotaxis
positive regulation of interleukin-1 beta secretion
positive regulation of inflammatory response
positive regulation of calcium-mediated signaling
positive chemotaxis
positive regulation of protein kinase B signaling
positive regulation of calcium ion transport
regulation of sensory perception of pain
cell chemotaxis
positive regulation of ERK1 and ERK2 cascade
response to cholesterol
cellular response to interferon-gamma
cellular response to interleukin-1
cellular response to tumor necrosis factor
cellular response to organic cyclic compound
granulocyte chemotaxis
positive regulation of natural killer cell chemotaxis
protein kinase activity
calcium-dependent protein kinase C activity
chemokine activity
heparin binding
phospholipase activator activity
kinase activity
CCR1 chemokine receptor binding
CCR5 chemokine receptor binding
chemoattractant activity
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0