Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DNMT1 SIRNA Assays ›
          Gene Symbol:
          Dnmt1
          Gene Name:
          DNA (cytosine-5-)-methyltransferase 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 21922515 - 21968495 on Build Rnor_6.0
          UniGene ID:
          Rn.6955
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 46874

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Dnmt1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_053354.3 NP_445806.3 2 176
          GenBank mRNA AB012214.1 - 2 160
          AF116344.1 - 2 214
          D64060.1 - 2 48
          DV214494.1 - 2 142

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_001199431, NM_010066, NM_001199433, NM_001199432, AF036009, and others.

          Panther Classification:

          Molecular Function -

          DNA metabolism protein DNA methyltransferase nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          DNA methylation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          brain development
          aging
          response to heat
          response to toxic substance
          response to organic substance
          response to ionizing radiation
          maintenance of DNA methylation
          response to lead ion
          DNA methylation on cytosine within a CG sequence
          regulation of gene expression
          positive regulation of gene expression
          response to activity
          gene silencing
          chromatin modification
          neuron differentiation
          response to caffeine
          response to nutrient levels
          response to estradiol
          response to lipopolysaccharide
          DNA methylation on cytosine
          response to vitamin A
          response to testosterone
          cellular response to platelet-derived growth factor stimulus
          response to antidepressant
          regulation of cell proliferation
          response to drug
          DNA hypermethylation
          response to ethanol
          negative regulation of transcription, DNA-templated
          S-adenosylhomocysteine metabolic process
          S-adenosylmethioninamine metabolic process
          S-adenosylmethionine metabolic process
          positive regulation of histone H3-K4 methylation
          negative regulation of histone H3-K9 methylation
          cellular response to amino acid stimulus
          cellular response to lead ion
          cellular response to transforming growth factor beta stimulus
          C-5 methylation of cytosine
          cellular response to nerve growth factor stimulus
          DNA binding
          chromatin binding
          double-stranded DNA binding
          RNA binding
          DNA (cytosine-5-)-methyltransferase activity
          protein binding
          transcription factor binding
          methyltransferase activity
          zinc ion binding
          methyl-CpG binding
          S-adenosylmethionine-dependent methyltransferase activity
          DNA-methyltransferase activity
          protein domain specific binding
          estrogen receptor binding
          histone deacetylase binding
          unmethylated CpG binding
          DNA (cytosine-5-)-methyltransferase activity, acting on CpG substrates

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0