Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AGTR2 SIRNA Assays ›
          Gene Symbol:
          Agtr2
          Gene Name:
          angiotensin II receptor, type 2
          Gene Aliases:
          AT2-R, AT2R, AT<sub>2</sub>R
          Chromosome Location:
          Chr.X: 119389477 - 119393845 on Build Rnor_6.0
          UniGene ID:
          Rn.162367
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 47855

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Agtr2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012494.3 NP_036626.1 1 1032
          XM_006257432.2 XP_006257494.1 3 1374
          XM_008773399.1 XP_008771621.1 3 1183
          GenBank mRNA BC161802.1 - 2 1277 9102491
          D16840.1 - 3 1171

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_007429, S67465, BC003811, L32840, U04828, and others.

          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          regulation of systemic arterial blood pressure by circulatory renin-angiotensin
          renin-angiotensin regulation of aldosterone production
          vasodilation by angiotensin involved in regulation of systemic arterial blood pressure
          brain renin-angiotensin system
          cellular sodium ion homeostasis
          inflammatory response
          cell surface receptor signaling pathway
          G-protein coupled receptor signaling pathway coupled to cGMP nucleotide second messenger
          positive regulation of cell proliferation
          response to organonitrogen compound
          negative regulation of heart rate
          negative regulation of norepinephrine secretion
          cerebellar cortex development
          neuron differentiation
          negative regulation of icosanoid secretion
          positive regulation of phosphoprotein phosphatase activity
          intracellular signal transduction
          regulation of metanephros size
          exploration behavior
          positive regulation of renal sodium excretion
          aldosterone secretion
          angiotensin-activated signaling pathway
          dopamine biosynthetic process
          regulation of transcription factor import into nucleus
          negative regulation of neuron apoptotic process
          positive regulation of transcription, DNA-templated
          positive regulation of vasodilation
          negative regulation of fibroblast proliferation
          positive regulation of cytokine secretion
          positive regulation of nitric-oxide synthase activity
          negative regulation of neurotrophin TRK receptor signaling pathway
          kidney morphogenesis
          cell growth involved in cardiac muscle cell development
          cellular response to dexamethasone stimulus
          positive regulation of metanephric glomerulus development
          positive regulation of branching involved in ureteric bud morphogenesis
          positive regulation of extrinsic apoptotic signaling pathway
          G-protein alpha-subunit binding
          angiotensin type II receptor activity
          protein binding
          transcription factor binding
          peptide hormone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0